Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03303704_pri
          Assay Name:
          hsa-mir-106a
          Stem-loop ID:
          hsa-mir-106a hsa-mir-18b
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.X: 134170198 - 134170278 [-] on Build GRCh38
          Stem-loop Location:
          Chr.X: 134170198 - 134170278 [-] on Build GRCh38 Chr.X: 134170041 - 134170111 [-] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-106a-5p hsa-miR-106a-3p hsa-miR-18b-3p hsa-miR-18b-5p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03303704_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          121

          Chromosome Location:

          Chr. X - 134170538 - 134170538 [+] on 134170538 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-106a-5p MIMAT0000103
          Human hsa-miR-106a-3p MIMAT0004517
          Human hsa-miR-18b-3p MIMAT0004751
          Human hsa-miR-18b-5p MIMAT0001412

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-106a
          Stem-loop Accession # MI0000113
          Stem-loop Sequence
          CCUUGGCCAUGUAAAAGUGCUUACAGUGCAGGUAGCUUUUUGAGAUCUACUGCAAUGUAAGCACUUCUUACAUUACCAUGG
          Chromosome Location Chr. X - 134170198 - 134170278 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000103
          miRBase ID: hsa-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. X - 134170198 - 134170278 [-] on Build GRCh38

          miRBase Accession #: MIMAT0004517
          miRBase ID: hsa-miR-106a-3p
          Mature miRNA Sequence: CUGCAAUGUAAGCACUUCUUAC
          Chromosome Location: Chr. X - 134170198 - 134170278 [-] on Build GRCh38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 188 bases
          Stem-loop ID hsa-mir-18b
          Stem-loop Accession # MI0001518
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCA
          Chromosome Location Chr. X - 134170041 - 134170111 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004751
          miRBase ID: hsa-miR-18b-3p
          Mature miRNA Sequence: UGCCCUAAAUGCCCCUUCUGGC
          Chromosome Location: Chr. X - 134170041 - 134170111 [-] on Build GRCh38

          miRBase Accession #: MIMAT0001412
          miRBase ID: hsa-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: Chr. X - 134170041 - 134170111 [-] on Build GRCh38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 355 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12567
          Ambion® Pre-miR™ miRNA Precursor : PM12567
          TaqMan™ MicroRNA Assay : 002169
          mirVana® miRNA inhibitor : MH12567
          mirVana® miRNA mimic : MC12567
          TaqMan™ Advanced miRNA Assay : 478225_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13092
          Ambion® Pre-miR™ miRNA Precursor : PM13092
          TaqMan™ MicroRNA Assay : 002170
          mirVana® miRNA inhibitor : MH13092
          mirVana® miRNA mimic : MC13092
          TaqMan™ Advanced miRNA Assay : 478623_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12677
          Ambion® Pre-miR™ miRNA Precursor : PM12677
          TaqMan™ MicroRNA Assay : 002310
          mirVana® miRNA inhibitor : MH12677
          mirVana® miRNA mimic : MC12677
          TaqMan™ Advanced miRNA Assay : 478734_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10466
          Ambion® Pre-miR™ miRNA Precursor : PM10466
          TaqMan™ MicroRNA Assay : 002217
          mirVana® miRNA inhibitor : MH10466
          mirVana® miRNA mimic : MC10466
          TaqMan™ Advanced miRNA Assay : 478584_mir


          miRBase Alias:

          Human: hsa-mir-18b (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline