Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10466
          Assay Name: hsa-miR-18b-5p
          miRBase Accession Number: MI0001518
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134170041 - 134170111 [-] on Build GRCh38
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Species: Human, Anolis carolinensis, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Xenopus laevis, Xenopus tropicalis
          Product Type: mirVana® miRNA mimic
          Assay ID MC10466

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          UAAGGUGCAUCUAGUGCAGUUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-18b-5p MIMAT0001412
          Anolis carolinensis aca-miR-18b-5p MIMAT0021819
          Cricetulus griseus cgr-miR-18b MIMAT0041171
          Dasypus novemcinctus dno-miR-18b-5p MIMAT0047513
          Guinea pig cpo-miR-18b-5p MIMAT0046861
          Horse eca-miR-18b MIMAT0013200
          View More Rows
          Monodelphis domestica mdo-miR-18b-5p MIMAT0028766
          Ophiophagus hannah oha-miR-18b-5p MIMAT0036770
          Pan troglodytes ptr-miR-18b MIMAT0008054
          Pig ssc-miR-18b MIMAT0020585
          Pongo pygmaeus ppy-miR-18b MIMAT0015731
          Rabbit ocu-miR-18b-5p MIMAT0048113
          Rhesus monkey mml-miR-18b MIMAT0006163
          Xenopus laevis xla-miR-18a-5p MIMAT0001349
          Xenopus tropicalis xtr-miR-18b MIMAT0003706

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-18b
          Stem-loop Accession # MI0001518
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCA
          Chromosome Location Chr. X - 134170041 - 134170111 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001412
          miRBase ID: hsa-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: Chr. X - 134170041 - 134170111 [-] on Build GRCh38

          Anolis carolinensis

          Stem-loop ID aca-mir-18b
          Stem-loop Accession # MI0018784
          Stem-loop Sequence
          GAAACAGCUUCCUGUGCUAAGGUGCAUCUAGUGCAGUUAGUGAAGUAGCUUAGAAUCUACUGCCCUGAAUGCUCCUUCUGGCAUAGGAGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021819
          miRBase ID: aca-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-18b
          Stem-loop Accession # MI0033101
          Stem-loop Sequence
          UCUUGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGACUCUACUGCCCUAAAUGCCCCUUCUCGCACAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041171
          miRBase ID: cgr-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-18b
          Stem-loop Accession # MI0038911
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAAACAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047513
          miRBase ID: dno-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-18b
          Stem-loop Accession # MI0038567
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046861
          miRBase ID: cpo-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-18b
          Stem-loop Accession # MI0012947
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013200
          miRBase ID: eca-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-18b
          Stem-loop Accession # MI0023939
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAUGUAGUUUAGUAUCUACUGCCCUAAAUGCUCCUUCCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028766
          miRBase ID: mdo-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-18b
          Stem-loop Accession # MI0031382
          Stem-loop Sequence
          UUCCUUGUGCUAAGGUGCAUCUAGUGCAGUUAGUGGACUAACUUACAAUCUACUGCCCUGAGUGCUCCUUCUGGCACACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036770
          miRBase ID: oha-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-18b
          Stem-loop Accession # MI0008562
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008054
          miRBase ID: ptr-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-18b
          Stem-loop Accession # MI0017985
          Stem-loop Sequence
          UCUCUUGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCACAGGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020585
          miRBase ID: ssc-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-18b
          Stem-loop Accession # MI0014796
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015731
          miRBase ID: ppy-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-18b
          Stem-loop Accession # MI0039227
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAAGCAACUUCGAAUCUACUGCCCUAAAUGCCCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048113
          miRBase ID: ocu-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-18b
          Stem-loop Accession # MI0007590
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006163
          miRBase ID: mml-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-18a-1
          Stem-loop Accession # MI0001454
          Stem-loop Sequence
          CCUUGUGUUAAGGUGCAUCUAGUGCAGUUAGUGACAUAGCGUAGCAUCUACUGCCCUAAAUGCUCCUUUUGGCACAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001349
          miRBase ID: xla-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Stem-loop ID xla-mir-18a-2
          Stem-loop Accession # MI0038277
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGACAUAGCGUAGCAUCUACUGCCCUAAAUGCUCCUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001349
          miRBase ID: xla-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-18b
          Stem-loop Accession # MI0004959
          Stem-loop Sequence
          CCUUGUGUUAAGGUGCAUCUAGUGCAGUUAGUGACAUAGUGUAGCAUCUACUGCCCUAAAUGCUCCUUUUGGCACAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003706
          miRBase ID: xtr-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303704_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10466
          Ambion® Pre-miR™ miRNA Precursor : PM10466
          TaqMan™ MicroRNA Assay : 002217
          mirVana® miRNA inhibitor : MH10466
          TaqMan™ Advanced miRNA Assay : 478584_mir


          miRBase Alias:

          Human: hsa-miR-18b (v17)
          Aeromonas caviae Ae398: aca-miR-18b (v18)
          Xenopus laevis: xla-miR-18 (v21)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline