Search
Search

| Assay Name: | hsa-miR-18b |
|---|---|
| miRBase Accession Number: | MI0001518 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAAGGUGCAUCUAGUGCAGUUAG |
| Species: | Human, Anolis carolinensis, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Xenopus laevis, Xenopus tropicalis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UAAGGUGCAUCUAGUGCAGUUAG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-18b-5p | MIMAT0001412 | |
| Anolis carolinensis | aca-miR-18b-5p | MIMAT0021819 | |
| Cricetulus griseus | cgr-miR-18b | MIMAT0041171 | |
| Dasypus novemcinctus | dno-miR-18b-5p | MIMAT0047513 | |
| Guinea pig | cpo-miR-18b-5p | MIMAT0046861 | |
| Horse | eca-miR-18b | MIMAT0013200 | |
| View More Rows | |||
| Monodelphis domestica | mdo-miR-18b-5p | MIMAT0028766 | |
| Ophiophagus hannah | oha-miR-18b-5p | MIMAT0036770 | |
| Pan troglodytes | ptr-miR-18b | MIMAT0008054 | |
| Pig | ssc-miR-18b | MIMAT0020585 | |
| Pongo pygmaeus | ppy-miR-18b | MIMAT0015731 | |
| Rabbit | ocu-miR-18b-5p | MIMAT0048113 | |
| Rhesus monkey | mml-miR-18b | MIMAT0006163 | |
| Xenopus laevis | xla-miR-18a-5p | MIMAT0001349 | |
| Xenopus tropicalis | xtr-miR-18b | MIMAT0003706 | |