Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478584_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-18b-5p
          Stem-loop Accession Number: MI0001518
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134170041 - 134170111 [-] on Build GRCh38
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Species: Human, Anolis carolinensis, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Xenopus laevis, Xenopus tropicalis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478584_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          UAAGGUGCAUCUAGUGCAGUUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-18b-5p MIMAT0001412
          Anolis carolinensis aca-miR-18b-5p MIMAT0021819
          Cricetulus griseus cgr-miR-18b MIMAT0041171
          Dasypus novemcinctus dno-miR-18b-5p MIMAT0047513
          Guinea pig cpo-miR-18b-5p MIMAT0046861
          Horse eca-miR-18b MIMAT0013200
          View More Rows
          Monodelphis domestica mdo-miR-18b-5p MIMAT0028766
          Ophiophagus hannah oha-miR-18b-5p MIMAT0036770
          Pan troglodytes ptr-miR-18b MIMAT0008054
          Pig ssc-miR-18b MIMAT0020585
          Pongo pygmaeus ppy-miR-18b MIMAT0015731
          Rabbit ocu-miR-18b-5p MIMAT0048113
          Rhesus monkey mml-miR-18b MIMAT0006163
          Xenopus laevis xla-miR-18a-5p MIMAT0001349
          Xenopus tropicalis xtr-miR-18b MIMAT0003706

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-18b
          Stem-loop Accession # MI0001518
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCA
          Chromosome Location Chr. X - 134170041 - 134170111 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001412
          miRBase ID: hsa-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: Chr. X - 134170041 - 134170111 [-] on Build GRCh38

          Anolis carolinensis

          Stem-loop ID aca-mir-18b
          Stem-loop Accession # MI0018784
          Stem-loop Sequence
          GAAACAGCUUCCUGUGCUAAGGUGCAUCUAGUGCAGUUAGUGAAGUAGCUUAGAAUCUACUGCCCUGAAUGCUCCUUCUGGCAUAGGAGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021819
          miRBase ID: aca-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-18b
          Stem-loop Accession # MI0033101
          Stem-loop Sequence
          UCUUGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGACUCUACUGCCCUAAAUGCCCCUUCUCGCACAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041171
          miRBase ID: cgr-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-18b
          Stem-loop Accession # MI0038911
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAAACAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047513
          miRBase ID: dno-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-18b
          Stem-loop Accession # MI0038567
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046861
          miRBase ID: cpo-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-18b
          Stem-loop Accession # MI0012947
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013200
          miRBase ID: eca-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-18b
          Stem-loop Accession # MI0023939
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAUGUAGUUUAGUAUCUACUGCCCUAAAUGCUCCUUCCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028766
          miRBase ID: mdo-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-18b
          Stem-loop Accession # MI0031382
          Stem-loop Sequence
          UUCCUUGUGCUAAGGUGCAUCUAGUGCAGUUAGUGGACUAACUUACAAUCUACUGCCCUGAGUGCUCCUUCUGGCACACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036770
          miRBase ID: oha-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-18b
          Stem-loop Accession # MI0008562
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008054
          miRBase ID: ptr-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-18b
          Stem-loop Accession # MI0017985
          Stem-loop Sequence
          UCUCUUGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCACAGGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020585
          miRBase ID: ssc-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-18b
          Stem-loop Accession # MI0014796
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015731
          miRBase ID: ppy-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-18b
          Stem-loop Accession # MI0039227
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGAAGCAACUUCGAAUCUACUGCCCUAAAUGCCCCUUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048113
          miRBase ID: ocu-miR-18b-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-18b
          Stem-loop Accession # MI0007590
          Stem-loop Sequence
          UGUGUUAAGGUGCAUCUAGUGCAGUUAGUGAAGCAGCUUAGAAUCUACUGCCCUAAAUGCCCCUUCUGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006163
          miRBase ID: mml-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-18a-1
          Stem-loop Accession # MI0001454
          Stem-loop Sequence
          CCUUGUGUUAAGGUGCAUCUAGUGCAGUUAGUGACAUAGCGUAGCAUCUACUGCCCUAAAUGCUCCUUUUGGCACAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001349
          miRBase ID: xla-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Stem-loop ID xla-mir-18a-2
          Stem-loop Accession # MI0038277
          Stem-loop Sequence
          UAAGGUGCAUCUAGUGCAGUUAGUGACAUAGCGUAGCAUCUACUGCCCUAAAUGCUCCUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001349
          miRBase ID: xla-miR-18a-5p
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-18b
          Stem-loop Accession # MI0004959
          Stem-loop Sequence
          CCUUGUGUUAAGGUGCAUCUAGUGCAGUUAGUGACAUAGUGUAGCAUCUACUGCCCUAAAUGCUCCUUUUGGCACAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003706
          miRBase ID: xtr-miR-18b
          Mature miRNA Sequence: UAAGGUGCAUCUAGUGCAGUUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303704_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10466
          Ambion® Pre-miR™ miRNA Precursor : PM10466
          TaqMan™ MicroRNA Assay : 002217
          mirVana® miRNA inhibitor : MH10466
          mirVana® miRNA mimic : MC10466
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A Card


          miRBase Alias:

          Xenopus laevis: xla-miR-18 (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline