Search
Search

| Assay Name: | hsa-miR-18b-5p |
|---|---|
| miRBase Accession Number: | MI0001518 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAAGGUGCAUCUAGUGCAGUUAG |
| Species: | Human, Anolis carolinensis, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Xenopus laevis, Xenopus tropicalis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UAAGGUGCAUCUAGUGCAGUUAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-18b-5p | MIMAT0001412 |
| Anolis carolinensis | aca-miR-18b-5p | MIMAT0021819 |
| Cricetulus griseus | cgr-miR-18b | MIMAT0041171 |
| Dasypus novemcinctus | dno-miR-18b-5p | MIMAT0047513 |
| Guinea pig | cpo-miR-18b-5p | MIMAT0046861 |
| Horse | eca-miR-18b | MIMAT0013200 |
| View More Rows | ||
| Monodelphis domestica | mdo-miR-18b-5p | MIMAT0028766 |
| Ophiophagus hannah | oha-miR-18b-5p | MIMAT0036770 |
| Pan troglodytes | ptr-miR-18b | MIMAT0008054 |
| Pig | ssc-miR-18b | MIMAT0020585 |
| Pongo pygmaeus | ppy-miR-18b | MIMAT0015731 |
| Rabbit | ocu-miR-18b-5p | MIMAT0048113 |
| Rhesus monkey | mml-miR-18b | MIMAT0006163 |
| Xenopus laevis | xla-miR-18a-5p | MIMAT0001349 |
| Xenopus tropicalis | xtr-miR-18b | MIMAT0003706 |