Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12567
          Assay Name: hsa-miR-106a-5p
          miRBase Accession Number: MI0000113
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134170198 - 134170278 [-] on Build GRCh38
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Species: Human, Chrysemys picta, Columba livia, Guinea pig, Pteropus alecto, Rabbit, Taeniopygia guttata
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12567
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          AAAAGUGCUUACAGUGCAGGUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-106a-5p MIMAT0000103
          Chrysemys picta cpi-miR-106a-5p MIMAT0037721
          Columba livia cli-miR-106-5p MIMAT0038487
          Guinea pig cpo-miR-106a-5p MIMAT0046955
          Pteropus alecto pal-miR-106a-5p MIMAT0040204
          Rabbit ocu-miR-106a-5p MIMAT0048195
          View More Rows
          Taeniopygia guttata tgu-miR-106-5p MIMAT0014590

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-106a
          Stem-loop Accession # MI0000113
          Stem-loop Sequence
          CCUUGGCCAUGUAAAAGUGCUUACAGUGCAGGUAGCUUUUUGAGAUCUACUGCAAUGUAAGCACUUCUUACAUUACCAUGG
          Chromosome Location Chr. X - 134170198 - 134170278 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000103
          miRBase ID: hsa-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. X - 134170198 - 134170278 [-] on Build GRCh38

          Chrysemys picta

          Stem-loop ID cpi-mir-106a
          Stem-loop Accession # MI0029451
          Stem-loop Sequence
          UGAGUUGUGCAAAAGUGCUUACAGUGCAGGUAGUGUGUUGGUAUCUACUGCAGUGGAAGCACUUCUAGCAUUACAAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037721
          miRBase ID: cpi-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-106
          Stem-loop Accession # MI0029951
          Stem-loop Sequence
          UUGAGUUGUGCAAAAGUGCUUACAGUGCAGGUAGAGUUUAGGAUCUACUGCAGUAUAAGCACUUCUGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038487
          miRBase ID: cli-miR-106-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-106a
          Stem-loop Accession # MI0038618
          Stem-loop Sequence
          AAAAGUGCUUACAGUGCAGGUAGCUUUUUAAGAUCUACUGCAAUGCAAGCACUUCUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046955
          miRBase ID: cpo-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-106a
          Stem-loop Accession # MI0032549
          Stem-loop Sequence
          AAAAGUGCUUACAGUGCAGGUAGCUUUUUGAGAUCUACUGCAAUGCAAGCACUUCUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040204
          miRBase ID: pal-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-106a
          Stem-loop Accession # MI0039272
          Stem-loop Sequence
          AAAAGUGCUUACAGUGCAGGUAGCUUUUUGAGAUCUACUGCAAUGCGAGCACUUCUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048195
          miRBase ID: ocu-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-106
          Stem-loop Accession # MI0013818
          Stem-loop Sequence
          CAAGUUGUGCAAAAGUGCUUACAGUGCAGGUAGAGUUCAGGAUCUACUGCAGUUUAAGCACUUCUGGCAUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014590
          miRBase ID: tgu-miR-106-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303704_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12567
          TaqMan™ MicroRNA Assay : 002169
          mirVana® miRNA inhibitor : MH12567
          mirVana® miRNA mimic : MC12567
          TaqMan™ Advanced miRNA Assay : 478225_mir


          miRBase Alias:

          Human: hsa-miR-106a (v17)
          Trabulsiella guamensis ATCC 49490: tgu-miR-106 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline