Search
Search

| Assay Name: | hsa-miR-106a |
|---|---|
| miRBase Accession Number: | MI0000113 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAAGUGCUUACAGUGCAGGUAG |
| Species: | Human, Chrysemys picta, Columba livia, Guinea pig, Pteropus alecto, Rabbit, Taeniopygia guttata |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | AAAAGUGCUUACAGUGCAGGUAG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-106a-5p | MIMAT0000103 | |
| Chrysemys picta | cpi-miR-106a-5p | MIMAT0037721 | |
| Columba livia | cli-miR-106-5p | MIMAT0038487 | |
| Guinea pig | cpo-miR-106a-5p | MIMAT0046955 | |
| Pteropus alecto | pal-miR-106a-5p | MIMAT0040204 | |
| Rabbit | ocu-miR-106a-5p | MIMAT0048195 | |
| View More Rows | |||
| Taeniopygia guttata | tgu-miR-106-5p | MIMAT0014590 | |