Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478225_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-106a-5p
          Stem-loop Accession Number: MI0000113
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134170198 - 134170278 [-] on Build GRCh38
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Species: Human, Chrysemys picta, Columba livia, Guinea pig, Pteropus alecto, Rabbit, Taeniopygia guttata
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478225_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          AAAAGUGCUUACAGUGCAGGUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-106a-5p MIMAT0000103
          Chrysemys picta cpi-miR-106a-5p MIMAT0037721
          Columba livia cli-miR-106-5p MIMAT0038487
          Guinea pig cpo-miR-106a-5p MIMAT0046955
          Pteropus alecto pal-miR-106a-5p MIMAT0040204
          Rabbit ocu-miR-106a-5p MIMAT0048195
          View More Rows
          Taeniopygia guttata tgu-miR-106-5p MIMAT0014590

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-106a
          Stem-loop Accession # MI0000113
          Stem-loop Sequence
          CCUUGGCCAUGUAAAAGUGCUUACAGUGCAGGUAGCUUUUUGAGAUCUACUGCAAUGUAAGCACUUCUUACAUUACCAUGG
          Chromosome Location Chr. X - 134170198 - 134170278 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000103
          miRBase ID: hsa-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. X - 134170198 - 134170278 [-] on Build GRCh38

          Chrysemys picta

          Stem-loop ID cpi-mir-106a
          Stem-loop Accession # MI0029451
          Stem-loop Sequence
          UGAGUUGUGCAAAAGUGCUUACAGUGCAGGUAGUGUGUUGGUAUCUACUGCAGUGGAAGCACUUCUAGCAUUACAAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037721
          miRBase ID: cpi-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-106
          Stem-loop Accession # MI0029951
          Stem-loop Sequence
          UUGAGUUGUGCAAAAGUGCUUACAGUGCAGGUAGAGUUUAGGAUCUACUGCAGUAUAAGCACUUCUGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038487
          miRBase ID: cli-miR-106-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-106a
          Stem-loop Accession # MI0038618
          Stem-loop Sequence
          AAAAGUGCUUACAGUGCAGGUAGCUUUUUAAGAUCUACUGCAAUGCAAGCACUUCUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046955
          miRBase ID: cpo-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-106a
          Stem-loop Accession # MI0032549
          Stem-loop Sequence
          AAAAGUGCUUACAGUGCAGGUAGCUUUUUGAGAUCUACUGCAAUGCAAGCACUUCUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040204
          miRBase ID: pal-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-106a
          Stem-loop Accession # MI0039272
          Stem-loop Sequence
          AAAAGUGCUUACAGUGCAGGUAGCUUUUUGAGAUCUACUGCAAUGCGAGCACUUCUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048195
          miRBase ID: ocu-miR-106a-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-106
          Stem-loop Accession # MI0013818
          Stem-loop Sequence
          CAAGUUGUGCAAAAGUGCUUACAGUGCAGGUAGAGUUCAGGAUCUACUGCAGUUUAAGCACUUCUGGCAUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014590
          miRBase ID: tgu-miR-106-5p
          Mature miRNA Sequence: AAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303704_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12567
          Ambion® Pre-miR™ miRNA Precursor : PM12567
          TaqMan™ MicroRNA Assay : 002169
          mirVana® miRNA inhibitor : MH12567
          mirVana® miRNA mimic : MC12567
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline