Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002368
          Assay Name: hsa-miR-497*
          miRBase Accession Number: MI0003138
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 7017911 - 7018022 [-] on Build GRCh38
          Mature miRNA Sequence: CAAACCACACUGUGGUGUUAGA
          Species: Human, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Monodelphis domestica, Rabbit
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002368
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000231, mir-497

          Mature miRNA Sequence:

          CAAACCACACUGUGGUGUUAGA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-497-3p MIMAT0004768
          Cricetulus griseus cgr-miR-497-3p MIMAT0023971
          Dasypus novemcinctus dno-miR-497-3p MIMAT0047935
          Guinea pig cpo-miR-497-3p MIMAT0047296
          Monodelphis domestica mdo-miR-497-3p MIMAT0026946
          Rabbit ocu-miR-497-3p MIMAT0048488

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-497
          Stem-loop Accession # MI0003138
          Stem-loop Sequence
          CCACCCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGG
          Chromosome Location Chr. 17 - 7017911 - 7018022 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004768
          miRBase ID: hsa-miR-497-3p
          Mature miRNA Sequence: CAAACCACACUGUGGUGUUAGA
          Chromosome Location: Chr. 17 - 7017911 - 7018022 [-] on Build GRCh38

          Cricetulus griseus

          Stem-loop ID cgr-mir-497
          Stem-loop Accession # MI0020532
          Stem-loop Sequence
          CUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023971
          miRBase ID: cgr-miR-497-3p
          Mature miRNA Sequence: CAAACCACACUGUGGUGUUAGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-497
          Stem-loop Accession # MI0039133
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGCACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047935
          miRBase ID: dno-miR-497-3p
          Mature miRNA Sequence: CAAACCACACUGUGGUGUUAGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-497
          Stem-loop Accession # MI0038797
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047296
          miRBase ID: cpo-miR-497-3p
          Mature miRNA Sequence: CAAACCACACUGUGGUGUUAGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-497
          Stem-loop Accession # MI0012519
          Stem-loop Sequence
          CCUGCCUCCGCCCCAGCAGCACACUGUGGUUUGUUCAGUGUUAUGGCCAAGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026946
          miRBase ID: mdo-miR-497-3p
          Mature miRNA Sequence: CAAACCACACUGUGGUGUUAGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-497
          Stem-loop Accession # MI0039426
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048488
          miRBase ID: ocu-miR-497-3p
          Mature miRNA Sequence: CAAACCACACUGUGGUGUUAGA
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303903_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12884
          Ambion® Pre-miR™ miRNA Precursor : PM12884
          mirVana® miRNA inhibitor : MH12884
          mirVana® miRNA mimic : MC12884
          TaqMan™ Advanced miRNA Assay : 478946_mir
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : TaqManTM Human MicroRNA Array B v3
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0


          miRBase Alias:

          Human: hsa-miR-497* (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-l699q:80/100.66.79.160:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline