Search
Search

| Assay Name: | mmu-miR-742* |
|---|---|
| miRBase Accession Number: | MI0005206 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACUCACAUGGUUGCUAAUCA |
| Species: | Mouse, Rat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000412, mir-742 |
Mature miRNA Sequence: | UACUCACAUGGUUGCUAAUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-742-5p | MIMAT0004838 |
| Rat | rno-miR-742-5p | MIMAT0017314 |