Search
Search

| Assay Name: | mmu-miR-145* |
|---|---|
| miRBase Accession Number: | MI0000169 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUUCCUGGAAAUACUGUUCUUG |
| Species: | Mouse |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000079, mir-145 |
Mature miRNA Sequence: | AUUCCUGGAAAUACUGUUCUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-145a-3p | MIMAT0004534 |