Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002584
          Assay Name: mmu-miR-34c*
          miRBase Accession Number: MI0000403
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 51103034 - 51103110 [-] on Build GRCm38
          Mature miRNA Sequence: AAUCACUAACCACACAGCCAGG
          Species: Mouse, Rat, Chicken
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002584
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AAUCACUAACCACACAGCCAGG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-34c-3p MIMAT0004580
          Rat rno-miR-34c-3p MIMAT0004723
          Chicken gga-miR-34c-3p MIMAT0026541

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-34c
          Stem-loop Accession # MI0000403
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACAGCCAGGUAAAAAGACU
          Chromosome Location Chr. 9 - 51103034 - 51103110 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004580
          miRBase ID: mmu-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACAGCCAGG
          Chromosome Location: Chr. 9 - 51103034 - 51103110 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-34c
          Stem-loop Accession # MI0000876
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACAGCCAGGUAAAAAGACU
          Chromosome Location Chr. 8 - 55492024 - 55492100 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004723
          miRBase ID: rno-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACAGCCAGG
          Chromosome Location: Chr. 8 - 55492024 - 55492100 [-] on Build Rnor_6.0

          Chicken

          Stem-loop ID gga-mir-34c
          Stem-loop Accession # MI0001261
          Stem-loop Sequence
          AGCCUGGUUACCAGGCAGUGUAGUUAGCUGAUUGCCACCAGGACCAAUCACUAACCACACAGCCAGGUAAAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026541
          miRBase ID: gga-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACAGCCAGG
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Rodent Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Rodent MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Rodent MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465278_pri
          TaqMan™ Pri-miRNA Assay : Mm03306660_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12245
          Ambion® Pre-miR™ miRNA Precursor : PM12245
          mirVana® miRNA inhibitor : MH12245
          mirVana® miRNA mimic : MC12245
          TaqMan™ Advanced miRNA Assay : mmu481802_mir
          TaqMan™ Advanced miRNA Assay : rno481085_mir
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v2.0
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v3.0
          MIRNA : TaqMan® Array Rodent MicroRNA B Cards v3.0
          FGMIR : TaqManTM Array Rodent MicroRNA B Card v3
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool B v3.0


          miRBase Alias:

          Mouse: mmu-miR-34c* (v17)
          Rat: rno-miR-34c* (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline