Search
Search

| Assay Name: | bta-miR-29a |
|---|---|
| miRBase Accession Number: | MI0004733 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAGCACCAUCUGAAAUCGGUUA |
| Species: | Bovine, Pig |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | CUAGCACCAUCUGAAAUCGGUUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-29a | MIMAT0003518 |
| Pig | ssc-miR-29a-3p | MIMAT0013870 |