Search
Search

| Assay Name: | bta-miR-2461-5p |
|---|---|
| miRBase Accession Number: | MI0011521 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCACUCUGUCUCGGAUCCUGGCA |
| Species: | Bovine |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CCACUCUGUCUCGGAUCCUGGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-2461-5p | MIMAT0012047 |