Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 245366_mat
          Assay Name: bta-miR-34c
          miRBase Accession Number: MI0018857
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Species: Alligator mississippiensis, Anolis carolinensis, Bovine, Chrysemys picta, Gorilla gorilla, Xenopus laevis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 245366_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AGGCAGUGUAGUUAGCUGAUUG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Alligator mississippiensis ami-miR-34b-5p MIMAT0038097
          Alligator mississippiensis ami-miR-34c-5p MIMAT0038099
          Anolis carolinensis aca-miR-34a-5p MIMAT0021939
          Anolis carolinensis aca-miR-34c-5p MIMAT0021943
          Bovine bta-miR-34c MIMAT0003854
          Chrysemys picta cpi-miR-34b-5p MIMAT0037694
          View More Rows
          Gorilla gorilla ggo-miR-34c MIMAT0024123
          Xenopus laevis xla-miR-34b-5p MIMAT0046586

          Stem-loop Details



          Alligator mississippiensis

          Stem-loop ID ami-mir-34b
          Stem-loop Accession # MI0029701
          Stem-loop Sequence
          AGUAGUGUUUGGUUUGUAGGCAGUGUAGUUAGCUGAUUGUGUCAGUCCCUUGGCAAUCACUAACUUCACUGCCAUCAAAACAAGGCACAGAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038097
          miRBase ID: ami-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Stem-loop ID ami-mir-34c
          Stem-loop Accession # MI0029702
          Stem-loop Sequence
          UACCUCAGCCUAGUUGCUAGGCAGUGUAGUUAGCUGAUUGCAAAAGGCAACAAUCACUAACCACACAGCCAGGUAAAAAGGUCUGUCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038099
          miRBase ID: ami-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-34a
          Stem-loop Accession # MI0018857
          Stem-loop Sequence
          UUGCCAGGCAGUGUAGUUAGCUGAUUGACGAGGCAACAGUCACUAACAACACGGCCAGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021939
          miRBase ID: aca-miR-34a-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Stem-loop ID aca-mir-34c
          Stem-loop Accession # MI0018859
          Stem-loop Sequence
          UGGUUUGUAGGCAGUGUAGUUAGCUGAUUGUGAGGUUUCCUUGGCAAUCACUAGCUUUACUGUCAUCAAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021943
          miRBase ID: aca-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-34c
          Stem-loop Accession # MI0005068
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAAUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003854
          miRBase ID: bta-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-34b
          Stem-loop Accession # MI0029435
          Stem-loop Sequence
          GAAGUAGUGUUUGGUUUGUAGGCAGUGUAGUUAGCUGAUUGUAUCCAUCUCUUUGCAAUCACUAACUUUACUGCCAUCAAAACAAGACACAGAAUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037694
          miRBase ID: cpi-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-34c
          Stem-loop Accession # MI0020670
          Stem-loop Sequence
          UUUCUAUGAGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUUUGGGAAUUCGUCCAAAUGAGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024123
          miRBase ID: ggo-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-34b
          Stem-loop Accession # MI0038326
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGUGUUAUAUCAAAUGUGCAAUCACUAACUAAACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046586
          miRBase ID: xla-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH11692
          mirVana® miRNA mimic : MC11692


          miRBase Alias:

          Aeromonas caviae Ae398: aca-miR-34c (v18)
          Aeromonas caviae Ae398: aca-miR-34a (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-nqctq:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0