Search
Search

| Assay Name: | aca-miR-34a-5p |
|---|---|
| miRBase Accession Number: | MI0018857 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCAGUGUAGUUAGCUGAUUG |
| Species: | Alligator mississippiensis, Anolis carolinensis, Bovine, Chrysemys picta, Gorilla gorilla, Xenopus laevis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000039, mir-34 |
Mature miRNA Sequence: | AGGCAGUGUAGUUAGCUGAUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-34b-5p | MIMAT0038097 |
| Alligator mississippiensis | ami-miR-34c-5p | MIMAT0038099 |
| Anolis carolinensis | aca-miR-34a-5p | MIMAT0021939 |
| Anolis carolinensis | aca-miR-34c-5p | MIMAT0021943 |
| Bovine | bta-miR-34c | MIMAT0003854 |
| Chrysemys picta | cpi-miR-34b-5p | MIMAT0037694 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-34c | MIMAT0024123 |
| Xenopus laevis | xla-miR-34b-5p | MIMAT0046586 |