Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11692
          Assay Name: aca-miR-34a-5p
          miRBase Accession Number: MI0018857
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Species: Alligator mississippiensis, Anolis carolinensis, Bovine, Chrysemys picta, Gorilla gorilla, Xenopus laevis
          Product Type: mirVana® miRNA mimic
          Assay ID MC11692

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AGGCAGUGUAGUUAGCUGAUUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Alligator mississippiensis ami-miR-34b-5p MIMAT0038097
          Alligator mississippiensis ami-miR-34c-5p MIMAT0038099
          Anolis carolinensis aca-miR-34a-5p MIMAT0021939
          Anolis carolinensis aca-miR-34c-5p MIMAT0021943
          Bovine bta-miR-34c MIMAT0003854
          Chrysemys picta cpi-miR-34b-5p MIMAT0037694
          View More Rows
          Gorilla gorilla ggo-miR-34c MIMAT0024123
          Xenopus laevis xla-miR-34b-5p MIMAT0046586

          Stem-loop Details



          Alligator mississippiensis

          Stem-loop ID ami-mir-34b
          Stem-loop Accession # MI0029701
          Stem-loop Sequence
          AGUAGUGUUUGGUUUGUAGGCAGUGUAGUUAGCUGAUUGUGUCAGUCCCUUGGCAAUCACUAACUUCACUGCCAUCAAAACAAGGCACAGAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038097
          miRBase ID: ami-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Stem-loop ID ami-mir-34c
          Stem-loop Accession # MI0029702
          Stem-loop Sequence
          UACCUCAGCCUAGUUGCUAGGCAGUGUAGUUAGCUGAUUGCAAAAGGCAACAAUCACUAACCACACAGCCAGGUAAAAAGGUCUGUCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038099
          miRBase ID: ami-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-34a
          Stem-loop Accession # MI0018857
          Stem-loop Sequence
          UUGCCAGGCAGUGUAGUUAGCUGAUUGACGAGGCAACAGUCACUAACAACACGGCCAGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021939
          miRBase ID: aca-miR-34a-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Stem-loop ID aca-mir-34c
          Stem-loop Accession # MI0018859
          Stem-loop Sequence
          UGGUUUGUAGGCAGUGUAGUUAGCUGAUUGUGAGGUUUCCUUGGCAAUCACUAGCUUUACUGUCAUCAAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021943
          miRBase ID: aca-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-34c
          Stem-loop Accession # MI0005068
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAAUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003854
          miRBase ID: bta-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-34b
          Stem-loop Accession # MI0029435
          Stem-loop Sequence
          GAAGUAGUGUUUGGUUUGUAGGCAGUGUAGUUAGCUGAUUGUAUCCAUCUCUUUGCAAUCACUAACUUUACUGCCAUCAAAACAAGACACAGAAUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037694
          miRBase ID: cpi-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-34c
          Stem-loop Accession # MI0020670
          Stem-loop Sequence
          UUUCUAUGAGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUUUGGGAAUUCGUCCAAAUGAGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024123
          miRBase ID: ggo-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-34b
          Stem-loop Accession # MI0038326
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGUGUUAUAUCAAAUGUGCAAUCACUAACUAAACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046586
          miRBase ID: xla-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 245366_mat
          mirVana® miRNA inhibitor : MH11692


          miRBase Alias:

          Aeromonas caviae Ae398: aca-miR-34c (v18)
          Aeromonas caviae Ae398: aca-miR-34a (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch