Search
Search

| Assay Name: | egr-miR-153 |
|---|---|
| miRBase Accession Number: | MI0017788 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCAUAGUCUCAUAAGUGCCA |
| Species: | Echinococcus granulosus, Echinococcus multilocularis, Mesocestoides corti |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | UUGCAUAGUCUCAUAAGUGCCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Echinococcus granulosus | egr-miR-153-3p | MIMAT0020244 |
| Echinococcus multilocularis | emu-miR-153-3p | MIMAT0020269 |
| Mesocestoides corti | mco-miR-153-5p | MIMAT0046794 |