Search
Search

| Assay Name: | efu-miR-660 |
|---|---|
| miRBase Accession Number: | MI0038832 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGUUGU |
| Species: | Dasypus novemcinctus, Eptesicus fuscus, Guinea pig, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGUUGU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Dasypus novemcinctus | dno-miR-660-5p | MIMAT0047994 | |
| Eptesicus fuscus | efu-miR-660 | MIMAT0035154 | |
| Guinea pig | cpo-miR-660-5p | MIMAT0047365 | |
| Microcebus murinus | mmr-miR-660 | MIMAT0049181 | |
| Nomascus leucogenys | nle-miR-660 | MIMAT0049361 | |
| Otolemur garnettii | oga-miR-660 | MIMAT0049689 | |
| View More Rows | |||
| Rabbit | ocu-miR-660-5p | MIMAT0048551 | |