Search
Search

| Assay Name: | cpo-miR-660-5p |
|---|---|
| miRBase Accession Number: | MI0038832 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGUUGU |
| Species: | Dasypus novemcinctus, Eptesicus fuscus, Guinea pig, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Rabbit |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGUUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dasypus novemcinctus | dno-miR-660-5p | MIMAT0047994 |
| Eptesicus fuscus | efu-miR-660 | MIMAT0035154 |
| Guinea pig | cpo-miR-660-5p | MIMAT0047365 |
| Microcebus murinus | mmr-miR-660 | MIMAT0049181 |
| Nomascus leucogenys | nle-miR-660 | MIMAT0049361 |
| Otolemur garnettii | oga-miR-660 | MIMAT0049689 |
| View More Rows | ||
| Rabbit | ocu-miR-660-5p | MIMAT0048551 |