Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH31497
          Assay Name: cpo-miR-660-5p
          miRBase Accession Number: MI0038832
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUGU
          Species: Dasypus novemcinctus, Eptesicus fuscus, Guinea pig, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Rabbit
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH31497

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Mature miRNA Sequence:

          UACCCAUUGCAUAUCGGAGUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Dasypus novemcinctus dno-miR-660-5p MIMAT0047994
          Eptesicus fuscus efu-miR-660 MIMAT0035154
          Guinea pig cpo-miR-660-5p MIMAT0047365
          Microcebus murinus mmr-miR-660 MIMAT0049181
          Nomascus leucogenys nle-miR-660 MIMAT0049361
          Otolemur garnettii oga-miR-660 MIMAT0049689
          View More Rows
          Rabbit ocu-miR-660-5p MIMAT0048551

          Stem-loop Details



          Dasypus novemcinctus

          Stem-loop ID dno-mir-660
          Stem-loop Accession # MI0039164
          Stem-loop Sequence
          UACCCAUUGCAUAUCGGAGUUGUGAUUUCUCAAAAACACCUCCUGUGUGCAUGGAUUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047994
          miRBase ID: dno-miR-660-5p
          Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUGU
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-660
          Stem-loop Accession # MI0028859
          Stem-loop Sequence
          CCUUCUCCCAUACCCAUUGCAUAUCGGAGUUGUGAAUUCUCAAAGCACCUCCUGUGUGCAUGGAUUACAGGAGGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035154
          miRBase ID: efu-miR-660
          Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-660
          Stem-loop Accession # MI0038832
          Stem-loop Sequence
          UACCCAUUGCAUAUCGGAGUUGUGAAUUCUUAAAAUAUCUCCUGUAUGCAUGGAUUAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047365
          miRBase ID: cpo-miR-660-5p
          Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-660
          Stem-loop Accession # MI0039899
          Stem-loop Sequence
          UACCCAUUGCAUAUCGGAGUUGUGAAUUCUCAAAACACCUCCUGUGUGCAUGGAUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049181
          miRBase ID: mmr-miR-660
          Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-660
          Stem-loop Accession # MI0040098
          Stem-loop Sequence
          UACCCAUUGCAUAUCGGAGUUGUGAAUUCUCAAAACACCUCCUGUGUGCAUGGAUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049361
          miRBase ID: nle-miR-660
          Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-660
          Stem-loop Accession # MI0040473
          Stem-loop Sequence
          UACCCAUUGCAUAUCGGAGUUGUGAAUUCUCAAAACACCUCCUGUGUGCAUGGAUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049689
          miRBase ID: oga-miR-660
          Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-660
          Stem-loop Accession # MI0039458
          Stem-loop Sequence
          UACCCAUUGCAUAUCGGAGUUGUAAAUUCUCAAAACACCUCCUGUGUGCAUGGAUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048551
          miRBase ID: ocu-miR-660-5p
          Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 475114_mat
          mirVana® miRNA mimic : MC31497

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline