Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478043_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-342-3p
          Stem-loop Accession Number: MI0000805
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100109655 - 100109753 [+] on Build GRCh38
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Species: Human, Mouse, Rat, Dog, Horse, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478043_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000190, mir-342

          Mature miRNA Sequence:

          UCUCACACAGAAAUCGCACCCGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-342-3p MIMAT0000753
          Mouse mmu-miR-342-3p MIMAT0000590
          Rat rno-miR-342-3p MIMAT0000589
          Dog cfa-miR-342 MIMAT0006709
          Horse eca-miR-342-3p MIMAT0013137
          Pan troglodytes ptr-miR-342 MIMAT0008110
          View More Rows
          Pongo pygmaeus ppy-miR-342-3p MIMAT0015845
          Pteropus alecto pal-miR-342-3p MIMAT0040086
          Rhesus monkey mml-miR-342-3p MIMAT0006283
          Tupaia chinensis tch-miR-342-3p MIMAT0036487

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-342
          Stem-loop Accession # MI0000805
          Stem-loop Sequence
          GAAACUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUUAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUACUUA
          Chromosome Location Chr. 14 - 100109655 - 100109753 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000753
          miRBase ID: hsa-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: Chr. 14 - 100109655 - 100109753 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-342
          Stem-loop Accession # MI0000627
          Stem-loop Sequence
          GAAAAUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUCAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUGCUGA
          Chromosome Location Chr. 12 - 108658620 - 108658718 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000590
          miRBase ID: mmu-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: Chr. 12 - 108658620 - 108658718 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-342
          Stem-loop Accession # MI0000626
          Stem-loop Sequence
          GAAAAUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUCAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUGCUGA
          Chromosome Location Chr. 6 - 132561169 - 132561267 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000589
          miRBase ID: rno-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: Chr. 6 - 132561169 - 132561267 [+] on Build Rnor_6.0

          Dog

          Stem-loop ID cfa-mir-342
          Stem-loop Accession # MI0008128
          Stem-loop Sequence
          GGGUGCUAUCUGUGAUUGAGGGACAUGGCAAAUAGAAUUGUCUCACACAGAAAUCGCACCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006709
          miRBase ID: cfa-miR-342
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-342
          Stem-loop Accession # MI0012884
          Stem-loop Sequence
          GAACCUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGCAAAUAGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCCGCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013137
          miRBase ID: eca-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-342
          Stem-loop Accession # MI0008629
          Stem-loop Sequence
          AAACUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGCUAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUACUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008110
          miRBase ID: ptr-miR-342
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-342
          Stem-loop Accession # MI0014905
          Stem-loop Sequence
          GAAACUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUUAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUACUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015845
          miRBase ID: ppy-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-342
          Stem-loop Accession # MI0032486
          Stem-loop Sequence
          AGGGGUGCUAUCUGUGAUUGAGGGACAUGGCAAACGGAAUUGUCUCACACAGAAAUCGCACCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040086
          miRBase ID: pal-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-342
          Stem-loop Accession # MI0007704
          Stem-loop Sequence
          GAAACUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUUAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUACUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006283
          miRBase ID: mml-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-342
          Stem-loop Accession # MI0031145
          Stem-loop Sequence
          AGGGGUGCUAUCUGUGACUGAGGGCAUGGUCAGCGGAUUGUCUCACACAGAAAUCGCACCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036487
          miRBase ID: tch-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307009_pri
          TaqMan™ Pri-miRNA Assay : Rn03464764_pri
          TaqMan™ Pri-miRNA Assay : Hs03303589_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12328
          Ambion® Pre-miR™ miRNA Precursor : PM12328
          TaqMan™ MicroRNA Assay : 002260
          mirVana® miRNA inhibitor : MH12328
          mirVana® miRNA mimic : MC12328
          TaqMan™ Advanced miRNA Assay : rno481074_mir
          TaqMan™ Advanced miRNA Assay : mmu481074_mir
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human A Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline