Search
Search

| Assay Name: | hsa-miR-342-3p |
|---|---|
| miRBase Accession Number: | MI0000805 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUCACACAGAAAUCGCACCCGU |
| Species: | Human, Mouse, Rat, Dog, Horse, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tupaia chinensis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000190, mir-342 |
Mature miRNA Sequence: | UCUCACACAGAAAUCGCACCCGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-342-3p | MIMAT0000753 |
| Mouse | mmu-miR-342-3p | MIMAT0000590 |
| Rat | rno-miR-342-3p | MIMAT0000589 |
| Dog | cfa-miR-342 | MIMAT0006709 |
| Horse | eca-miR-342-3p | MIMAT0013137 |
| Pan troglodytes | ptr-miR-342 | MIMAT0008110 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-342-3p | MIMAT0015845 |
| Pteropus alecto | pal-miR-342-3p | MIMAT0040086 |
| Rhesus monkey | mml-miR-342-3p | MIMAT0006283 |
| Tupaia chinensis | tch-miR-342-3p | MIMAT0036487 |