Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481074_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-342-3p
          Stem-loop Accession Number: MI0000805
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100109655 - 100109753 [+] on Build GRCh38
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Species: Human, Mouse, Rat, Dog, Horse, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481074_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000190, mir-342

          Mature miRNA Sequence:

          UCUCACACAGAAAUCGCACCCGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-342-3p MIMAT0000753
          Mouse mmu-miR-342-3p MIMAT0000590
          Rat rno-miR-342-3p MIMAT0000589
          Dog cfa-miR-342 MIMAT0006709
          Horse eca-miR-342-3p MIMAT0013137
          Pan troglodytes ptr-miR-342 MIMAT0008110
          View More Rows
          Pongo pygmaeus ppy-miR-342-3p MIMAT0015845
          Pteropus alecto pal-miR-342-3p MIMAT0040086
          Rhesus monkey mml-miR-342-3p MIMAT0006283
          Tupaia chinensis tch-miR-342-3p MIMAT0036487

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-342
          Stem-loop Accession # MI0000805
          Stem-loop Sequence
          GAAACUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUUAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUACUUA
          Chromosome Location Chr. 14 - 100109655 - 100109753 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000753
          miRBase ID: hsa-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: Chr. 14 - 100109655 - 100109753 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-342
          Stem-loop Accession # MI0000627
          Stem-loop Sequence
          GAAAAUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUCAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUGCUGA
          Chromosome Location Chr. 12 - 108658620 - 108658718 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000590
          miRBase ID: mmu-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: Chr. 12 - 108658620 - 108658718 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-342
          Stem-loop Accession # MI0000626
          Stem-loop Sequence
          GAAAAUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUCAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUGCUGA
          Chromosome Location Chr. 6 - 132561169 - 132561267 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000589
          miRBase ID: rno-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: Chr. 6 - 132561169 - 132561267 [+] on Build Rnor_6.0

          Dog

          Stem-loop ID cfa-mir-342
          Stem-loop Accession # MI0008128
          Stem-loop Sequence
          GGGUGCUAUCUGUGAUUGAGGGACAUGGCAAAUAGAAUUGUCUCACACAGAAAUCGCACCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006709
          miRBase ID: cfa-miR-342
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-342
          Stem-loop Accession # MI0012884
          Stem-loop Sequence
          GAACCUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGCAAAUAGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCCGCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013137
          miRBase ID: eca-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-342
          Stem-loop Accession # MI0008629
          Stem-loop Sequence
          AAACUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGCUAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUACUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008110
          miRBase ID: ptr-miR-342
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-342
          Stem-loop Accession # MI0014905
          Stem-loop Sequence
          GAAACUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUUAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUACUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015845
          miRBase ID: ppy-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-342
          Stem-loop Accession # MI0032486
          Stem-loop Sequence
          AGGGGUGCUAUCUGUGAUUGAGGGACAUGGCAAACGGAAUUGUCUCACACAGAAAUCGCACCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040086
          miRBase ID: pal-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-342
          Stem-loop Accession # MI0007704
          Stem-loop Sequence
          GAAACUGGGCUCAAGGUGAGGGGUGCUAUCUGUGAUUGAGGGACAUGGUUAAUGGAAUUGUCUCACACAGAAAUCGCACCCGUCACCUUGGCCUACUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006283
          miRBase ID: mml-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-342
          Stem-loop Accession # MI0031145
          Stem-loop Sequence
          AGGGGUGCUAUCUGUGACUGAGGGCAUGGUCAGCGGAUUGUCUCACACAGAAAUCGCACCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036487
          miRBase ID: tch-miR-342-3p
          Mature miRNA Sequence: UCUCACACAGAAAUCGCACCCGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307009_pri
          TaqMan™ Pri-miRNA Assay : Rn03464764_pri
          TaqMan™ Pri-miRNA Assay : Hs03303589_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12328
          Ambion® Pre-miR™ miRNA Precursor : PM12328
          TaqMan™ MicroRNA Assay : 002260
          mirVana® miRNA inhibitor : MH12328
          mirVana® miRNA mimic : MC12328
          TaqMan™ Advanced miRNA Assay : rno481074_mir
          TaqMan™ Advanced miRNA Assay : 478043_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline