Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478312_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-139-5p
          Stem-loop Accession Number: MI0000261
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 72615063 - 72615130 [-] on Build GRCh38
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Species: Human, Bovine, Callithrix jacchus, Dasypus novemcinctus, Rabbit, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478312_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000117, mir-139

          Mature miRNA Sequence:

          UCUACAGUGCACGUGUCUCCAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-139-5p MIMAT0000250
          Bovine bta-miR-139 MIMAT0003788
          Callithrix jacchus cja-miR-139 MIMAT0039472
          Dasypus novemcinctus dno-miR-139-5p MIMAT0047651
          Rabbit ocu-miR-139-5p MIMAT0048239
          Tupaia chinensis tch-miR-139-5p MIMAT0036540

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-139
          Stem-loop Accession # MI0000261
          Stem-loop Sequence
          GUGUAUUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAAC
          Chromosome Location Chr. 11 - 72615063 - 72615130 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000250
          miRBase ID: hsa-miR-139-5p
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: Chr. 11 - 72615063 - 72615130 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-139
          Stem-loop Accession # MI0005009
          Stem-loop Sequence
          GUGUACUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003788
          miRBase ID: bta-miR-139
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-139
          Stem-loop Accession # MI0031996
          Stem-loop Sequence
          CUGGCUCAGGUGUAUUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAACAACUGAAGCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039472
          miRBase ID: cja-miR-139
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-139
          Stem-loop Accession # MI0038988
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047651
          miRBase ID: dno-miR-139-5p
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-139
          Stem-loop Accession # MI0039297
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUUGGAGGCUGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048239
          miRBase ID: ocu-miR-139-5p
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-139
          Stem-loop Accession # MI0031199
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUCAGAGGCUGGAGACGCGGCCCUGUCGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036540
          miRBase ID: tch-miR-139-5p
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302845_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11749
          Ambion® Pre-miR™ miRNA Precursor : PM11749
          TaqMan™ MicroRNA Assay : 005364_mat
          mirVana® miRNA inhibitor : MH11749
          mirVana® miRNA mimic : MC11749
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human A Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline