Search
Search

| Assay Name: | hsa-miR-139-5p |
|---|---|
| miRBase Accession Number: | MI0000261 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUACAGUGCACGUGUCUCCAGU |
| Species: | Human, Bovine, Callithrix jacchus, Dasypus novemcinctus, Rabbit, Tupaia chinensis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000117, mir-139 |
Mature miRNA Sequence: | UCUACAGUGCACGUGUCUCCAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-139-5p | MIMAT0000250 |
| Bovine | bta-miR-139 | MIMAT0003788 |
| Callithrix jacchus | cja-miR-139 | MIMAT0039472 |
| Dasypus novemcinctus | dno-miR-139-5p | MIMAT0047651 |
| Rabbit | ocu-miR-139-5p | MIMAT0048239 |
| Tupaia chinensis | tch-miR-139-5p | MIMAT0036540 |