Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478494_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-10b-5p
          Stem-loop Accession Number: MI0000267
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 176150303 - 176150412 [+] on Build GRCh38
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Species: Human, Mouse, Bovine, Fugu rubripes, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Tetraodon nigroviridis, Tupaia chinensis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478494_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          UACCCUGUAGAACCGAAUUUGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-10b-5p MIMAT0000254
          Mouse mmu-miR-10b-5p MIMAT0000208
          Bovine bta-miR-10b MIMAT0003839
          Fugu rubripes fru-miR-10b MIMAT0002963
          Horse eca-miR-10b MIMAT0013090
          Pan troglodytes ptr-miR-10b MIMAT0007945
          View More Rows
          Pongo pygmaeus ppy-miR-10b MIMAT0015730
          Rhesus monkey mml-miR-10b-5p MIMAT0006162
          Tetraodon nigroviridis tni-miR-10b MIMAT0002964
          Tupaia chinensis tch-miR-10b-5p MIMAT0036471
          Zebrafish dre-miR-10b-5p MIMAT0001268

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-10b
          Stem-loop Accession # MI0000267
          Stem-loop Sequence
          CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location Chr. 2 - 176150303 - 176150412 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000254
          miRBase ID: hsa-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: Chr. 2 - 176150303 - 176150412 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-10b
          Stem-loop Accession # MI0000221
          Stem-loop Sequence
          UAUAUACCCUGUAGAACCGAAUUUGUGUGGUACCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUA
          Chromosome Location Chr. 2 - 74726070 - 74726137 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000208
          miRBase ID: mmu-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: Chr. 2 - 74726070 - 74726137 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-10b
          Stem-loop Accession # MI0005052
          Stem-loop Sequence
          CAGUGACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003839
          miRBase ID: bta-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-10b-1
          Stem-loop Accession # MI0003279
          Stem-loop Sequence
          AUAUAUACCCUGUAGAACCGAAUUUGUGUGAUGGCGUCAAAGUCACAGAUUCGAUUCUAGGGGAGUAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002963
          miRBase ID: fru-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Stem-loop ID fru-mir-10b-2
          Stem-loop Accession # MI0003297
          Stem-loop Sequence
          GUUGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUGAGUUCCAGACAGUCGCAAGUACGUCUCUACAGGAAUACAUGGGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002963
          miRBase ID: fru-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-10b
          Stem-loop Accession # MI0012837
          Stem-loop Sequence
          UAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013090
          miRBase ID: eca-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-10b
          Stem-loop Accession # MI0008411
          Stem-loop Sequence
          CAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007945
          miRBase ID: ptr-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-10b
          Stem-loop Accession # MI0014794
          Stem-loop Sequence
          CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015730
          miRBase ID: ppy-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-10b
          Stem-loop Accession # MI0007588
          Stem-loop Sequence
          CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006162
          miRBase ID: mml-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-10b-1
          Stem-loop Accession # MI0003298
          Stem-loop Sequence
          GUUGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUGAGUUCAGACAGUCACAAGUACGUCUCUACAGGAAUACAUGGGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002964
          miRBase ID: tni-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Stem-loop ID tni-mir-10b-2
          Stem-loop Accession # MI0003280
          Stem-loop Sequence
          AUAUAUACCCUGUAGAACCGAAUUUGUGUGAUCAAGUCACAGUCACAGAUUCGAUUCUAGGGGAGUAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002964
          miRBase ID: tni-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-10b-1
          Stem-loop Accession # MI0031130
          Stem-loop Sequence
          UACCCUGUAGAACCGAAUUUGUGUGGUAUCCGCAUAGUCACAGAUUCGAUUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036471
          miRBase ID: tch-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Stem-loop ID tch-mir-10b-2
          Stem-loop Accession # MI0031249
          Stem-loop Sequence
          UACCCUGUAGAACCGAAUUUGUGUGGUAUCCGCAUAGUCACAGAUUCGAUUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036471
          miRBase ID: tch-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-10b-1
          Stem-loop Accession # MI0001364
          Stem-loop Sequence
          GUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAAAAAAUAACAUUCACAGAUUCGAUUCUAGGGGAGUAUAUGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001268
          miRBase ID: dre-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Stem-loop ID dre-mir-10b-2
          Stem-loop Accession # MI0001887
          Stem-loop Sequence
          GUAGUCGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUCCAAAACAUCAAAAUCGCAAAUACGUCUCUACAGGAAUACAUGGGCGACGUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001268
          miRBase ID: dre-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306401_pri
          TaqMan™ Pri-miRNA Assay : Hs03302879_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11108
          Ambion® Pre-miR™ miRNA Precursor : PM11108
          TaqMan™ MicroRNA Assay : 002218
          mirVana® miRNA inhibitor : MH11108
          mirVana® miRNA mimic : MC11108
          TaqMan™ Advanced miRNA Assay : mmu478494_mir
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline