Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11108
          Assay Name: hsa-miR-10b-5p
          miRBase Accession Number: MI0000267
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 176150303 - 176150412 [+] on Build GRCh38
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Species: Human, Mouse, Bovine, Fugu rubripes, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Tetraodon nigroviridis, Tupaia chinensis, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC11108

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          UACCCUGUAGAACCGAAUUUGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-10b-5p MIMAT0000254
          Mouse mmu-miR-10b-5p MIMAT0000208
          Bovine bta-miR-10b MIMAT0003839
          Fugu rubripes fru-miR-10b MIMAT0002963
          Horse eca-miR-10b MIMAT0013090
          Pan troglodytes ptr-miR-10b MIMAT0007945
          View More Rows
          Pongo pygmaeus ppy-miR-10b MIMAT0015730
          Rhesus monkey mml-miR-10b-5p MIMAT0006162
          Tetraodon nigroviridis tni-miR-10b MIMAT0002964
          Tupaia chinensis tch-miR-10b-5p MIMAT0036471
          Zebrafish dre-miR-10b-5p MIMAT0001268

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-10b
          Stem-loop Accession # MI0000267
          Stem-loop Sequence
          CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location Chr. 2 - 176150303 - 176150412 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000254
          miRBase ID: hsa-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: Chr. 2 - 176150303 - 176150412 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-10b
          Stem-loop Accession # MI0000221
          Stem-loop Sequence
          UAUAUACCCUGUAGAACCGAAUUUGUGUGGUACCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUA
          Chromosome Location Chr. 2 - 74726070 - 74726137 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000208
          miRBase ID: mmu-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: Chr. 2 - 74726070 - 74726137 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-10b
          Stem-loop Accession # MI0005052
          Stem-loop Sequence
          CAGUGACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003839
          miRBase ID: bta-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-10b-1
          Stem-loop Accession # MI0003279
          Stem-loop Sequence
          AUAUAUACCCUGUAGAACCGAAUUUGUGUGAUGGCGUCAAAGUCACAGAUUCGAUUCUAGGGGAGUAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002963
          miRBase ID: fru-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Stem-loop ID fru-mir-10b-2
          Stem-loop Accession # MI0003297
          Stem-loop Sequence
          GUUGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUGAGUUCCAGACAGUCGCAAGUACGUCUCUACAGGAAUACAUGGGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002963
          miRBase ID: fru-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-10b
          Stem-loop Accession # MI0012837
          Stem-loop Sequence
          UAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013090
          miRBase ID: eca-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-10b
          Stem-loop Accession # MI0008411
          Stem-loop Sequence
          CAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007945
          miRBase ID: ptr-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-10b
          Stem-loop Accession # MI0014794
          Stem-loop Sequence
          CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015730
          miRBase ID: ppy-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-10b
          Stem-loop Accession # MI0007588
          Stem-loop Sequence
          CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCAUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006162
          miRBase ID: mml-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-10b-1
          Stem-loop Accession # MI0003298
          Stem-loop Sequence
          GUUGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUGAGUUCAGACAGUCACAAGUACGUCUCUACAGGAAUACAUGGGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002964
          miRBase ID: tni-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Stem-loop ID tni-mir-10b-2
          Stem-loop Accession # MI0003280
          Stem-loop Sequence
          AUAUAUACCCUGUAGAACCGAAUUUGUGUGAUCAAGUCACAGUCACAGAUUCGAUUCUAGGGGAGUAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002964
          miRBase ID: tni-miR-10b
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-10b-1
          Stem-loop Accession # MI0031130
          Stem-loop Sequence
          UACCCUGUAGAACCGAAUUUGUGUGGUAUCCGCAUAGUCACAGAUUCGAUUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036471
          miRBase ID: tch-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Stem-loop ID tch-mir-10b-2
          Stem-loop Accession # MI0031249
          Stem-loop Sequence
          UACCCUGUAGAACCGAAUUUGUGUGGUAUCCGCAUAGUCACAGAUUCGAUUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036471
          miRBase ID: tch-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-10b-1
          Stem-loop Accession # MI0001364
          Stem-loop Sequence
          GUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGAAAAAAUAACAUUCACAGAUUCGAUUCUAGGGGAGUAUAUGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001268
          miRBase ID: dre-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA

          Stem-loop ID dre-mir-10b-2
          Stem-loop Accession # MI0001887
          Stem-loop Sequence
          GUAGUCGUCUAUAUGUACCCUGUAGAACCGAAUUUGUGUCCAAAACAUCAAAAUCGCAAAUACGUCUCUACAGGAAUACAUGGGCGACGUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001268
          miRBase ID: dre-miR-10b-5p
          Mature miRNA Sequence: UACCCUGUAGAACCGAAUUUGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306401_pri
          TaqMan™ Pri-miRNA Assay : Hs03302879_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11108
          Ambion® Pre-miR™ miRNA Precursor : PM11108
          TaqMan™ MicroRNA Assay : 002218
          mirVana® miRNA inhibitor : MH11108
          TaqMan™ Advanced miRNA Assay : mmu478494_mir
          TaqMan™ Advanced miRNA Assay : 478494_mir


          miRBase Alias:

          Human: hsa-miR-10b (v17)
          Mouse: mmu-miR-10b (v17)
          Danio rerio: dre-miR-10b (v20)
          Macaca mulatta: mml-miR-10b (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline