Search
Search

| Assay Name: | mmu-miR-10b-5p |
|---|---|
| Stem-loop Accession Number: | MI0000267 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACCCUGUAGAACCGAAUUUGUG |
| Species: | Human, Mouse, Bovine, Fugu rubripes, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Tetraodon nigroviridis, Tupaia chinensis, Zebrafish |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | UACCCUGUAGAACCGAAUUUGUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-10b-5p | MIMAT0000254 |
| Mouse | mmu-miR-10b-5p | MIMAT0000208 |
| Bovine | bta-miR-10b | MIMAT0003839 |
| Fugu rubripes | fru-miR-10b | MIMAT0002963 |
| Horse | eca-miR-10b | MIMAT0013090 |
| Pan troglodytes | ptr-miR-10b | MIMAT0007945 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-10b | MIMAT0015730 |
| Rhesus monkey | mml-miR-10b-5p | MIMAT0006162 |
| Tetraodon nigroviridis | tni-miR-10b | MIMAT0002964 |
| Tupaia chinensis | tch-miR-10b-5p | MIMAT0036471 |
| Zebrafish | dre-miR-10b-5p | MIMAT0001268 |