Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478717_mir

          Assay Name:
          Stem-loop Accession Number: MI0005482
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 122640803 - 122640881 [+] on Build GRCm38
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Species: Mouse, Rat, Bovine, Columba livia, Dog, Pan troglodytes, Pongo pygmaeus, Python bivittatus
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478717_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000105, mir-147

          Mature miRNA Sequence:

          GUGUGCGGAAAUGCUUCUGCUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-147-3p MIMAT0004857
          Rat rno-miR-147 MIMAT0005297
          Bovine bta-miR-147 MIMAT0009237
          Columba livia cli-miR-147-3p MIMAT0038541
          Dog cfa-miR-147 MIMAT0009874
          Pan troglodytes ptr-miR-147b MIMAT0008040
          View More Rows
          Pongo pygmaeus ppy-miR-147b MIMAT0015770
          Python bivittatus pbv-miR-147-3p MIMAT0038963

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-147
          Stem-loop Accession # MI0005482
          Stem-loop Sequence
          UAUGAAUCUAGUGGAAACAUUUCUGCACAAACUAGAUGUUGAUGCCAGUGUGCGGAAAUGCUUCUGCUACAUUUGUAGG
          Chromosome Location Chr. 2 - 122640803 - 122640881 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004857
          miRBase ID: mmu-miR-147-3p
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: Chr. 2 - 122640803 - 122640881 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-147
          Stem-loop Accession # MI0006130
          Stem-loop Sequence
          UAUGAAUCUAGUGGAAACACUUCUGCACAAACUCGAUUUUGAUGCCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGG
          Chromosome Location Chr. 3 - 114775343 - 114775421 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005297
          miRBase ID: rno-miR-147
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: Chr. 3 - 114775343 - 114775421 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-147
          Stem-loop Accession # MI0009747
          Stem-loop Sequence
          UAUGAAUCUAGUGGAAACACUUCUGCACAGGCUAGAUUAUGGAUACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009237
          miRBase ID: bta-miR-147
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-147
          Stem-loop Accession # MI0029983
          Stem-loop Sequence
          GGUACCCUAUGAAUCUAGUGGAAUCACUUCUGCACAAACUUGACUACUGAAAUCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGGGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038541
          miRBase ID: cli-miR-147-3p
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-147
          Stem-loop Accession # MI0010371
          Stem-loop Sequence
          ACUCUAUGAAUCUAGUGGAAACAUUGCUGCACAAACUAGAUUAUGGAUACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009874
          miRBase ID: cfa-miR-147
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-147b
          Stem-loop Accession # MI0008546
          Stem-loop Sequence
          AUAAAUCUAGUGGAAACAUUUCUGCACAAACUAGAUUCUGGACACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008040
          miRBase ID: ptr-miR-147b
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-147b
          Stem-loop Accession # MI0014836
          Stem-loop Sequence
          UAUAAAUCUAGUGGAAACAUUUCUGCACAAACUAGAUUCUGGACACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015770
          miRBase ID: ppy-miR-147b
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-147
          Stem-loop Accession # MI0030246
          Stem-loop Sequence
          CACUCUAUGAAUCUAGUGGAAUCACUUCUGCACAAACUAGAGGAUUGAAACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGAGUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038963
          miRBase ID: pbv-miR-147-3p
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464693_pri
          TaqMan™ Pri-miRNA Assay : Mm03308429_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12874
          Ambion® Pre-miR™ miRNA Precursor : PM12874
          TaqMan™ MicroRNA Assay : 002262
          mirVana® miRNA inhibitor : MH12874
          mirVana® miRNA mimic : MC12874
          TaqMan™ Advanced miRNA Assay : rno480942_mir
          TaqMan™ Advanced miRNA Assay : mmu482614_mir
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1


          miRBase Alias:

          Human: hsa-miR-147b (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline