Search
Search

| Assay Name: | mmu-miR-147-3p |
|---|---|
| miRBase Accession Number: | MI0005482 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUGUGCGGAAAUGCUUCUGCUA |
| Species: | Mouse, Rat, Bovine, Columba livia, Dog, Pan troglodytes, Pongo pygmaeus, Python bivittatus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000105, mir-147 |
Mature miRNA Sequence: | GUGUGCGGAAAUGCUUCUGCUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-147-3p | MIMAT0004857 |
| Rat | rno-miR-147 | MIMAT0005297 |
| Bovine | bta-miR-147 | MIMAT0009237 |
| Columba livia | cli-miR-147-3p | MIMAT0038541 |
| Dog | cfa-miR-147 | MIMAT0009874 |
| Pan troglodytes | ptr-miR-147b | MIMAT0008040 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-147b | MIMAT0015770 |
| Python bivittatus | pbv-miR-147-3p | MIMAT0038963 |