Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12874
          Assay Name: mmu-miR-147-3p
          miRBase Accession Number: MI0005482
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 122640803 - 122640881 [+] on Build GRCm38
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Species: Mouse, Rat, Bovine, Columba livia, Dog, Pan troglodytes, Pongo pygmaeus, Python bivittatus
          Product Type: mirVana® miRNA mimic
          Assay ID MC12874

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000105, mir-147

          Mature miRNA Sequence:

          GUGUGCGGAAAUGCUUCUGCUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-147-3p MIMAT0004857
          Rat rno-miR-147 MIMAT0005297
          Bovine bta-miR-147 MIMAT0009237
          Columba livia cli-miR-147-3p MIMAT0038541
          Dog cfa-miR-147 MIMAT0009874
          Pan troglodytes ptr-miR-147b MIMAT0008040
          View More Rows
          Pongo pygmaeus ppy-miR-147b MIMAT0015770
          Python bivittatus pbv-miR-147-3p MIMAT0038963

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-147
          Stem-loop Accession # MI0005482
          Stem-loop Sequence
          UAUGAAUCUAGUGGAAACAUUUCUGCACAAACUAGAUGUUGAUGCCAGUGUGCGGAAAUGCUUCUGCUACAUUUGUAGG
          Chromosome Location Chr. 2 - 122640803 - 122640881 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004857
          miRBase ID: mmu-miR-147-3p
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: Chr. 2 - 122640803 - 122640881 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-147
          Stem-loop Accession # MI0006130
          Stem-loop Sequence
          UAUGAAUCUAGUGGAAACACUUCUGCACAAACUCGAUUUUGAUGCCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGG
          Chromosome Location Chr. 3 - 114775343 - 114775421 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005297
          miRBase ID: rno-miR-147
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: Chr. 3 - 114775343 - 114775421 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-147
          Stem-loop Accession # MI0009747
          Stem-loop Sequence
          UAUGAAUCUAGUGGAAACACUUCUGCACAGGCUAGAUUAUGGAUACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009237
          miRBase ID: bta-miR-147
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-147
          Stem-loop Accession # MI0029983
          Stem-loop Sequence
          GGUACCCUAUGAAUCUAGUGGAAUCACUUCUGCACAAACUUGACUACUGAAAUCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGGGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038541
          miRBase ID: cli-miR-147-3p
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-147
          Stem-loop Accession # MI0010371
          Stem-loop Sequence
          ACUCUAUGAAUCUAGUGGAAACAUUGCUGCACAAACUAGAUUAUGGAUACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009874
          miRBase ID: cfa-miR-147
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-147b
          Stem-loop Accession # MI0008546
          Stem-loop Sequence
          AUAAAUCUAGUGGAAACAUUUCUGCACAAACUAGAUUCUGGACACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008040
          miRBase ID: ptr-miR-147b
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-147b
          Stem-loop Accession # MI0014836
          Stem-loop Sequence
          UAUAAAUCUAGUGGAAACAUUUCUGCACAAACUAGAUUCUGGACACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015770
          miRBase ID: ppy-miR-147b
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-147
          Stem-loop Accession # MI0030246
          Stem-loop Sequence
          CACUCUAUGAAUCUAGUGGAAUCACUUCUGCACAAACUAGAGGAUUGAAACCAGUGUGCGGAAAUGCUUCUGCUACAUUUUUAGAGUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038963
          miRBase ID: pbv-miR-147-3p
          Mature miRNA Sequence: GUGUGCGGAAAUGCUUCUGCUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464693_pri
          TaqMan™ Pri-miRNA Assay : Mm03308429_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12874
          Ambion® Pre-miR™ miRNA Precursor : PM12874
          TaqMan™ MicroRNA Assay : 002262
          mirVana® miRNA inhibitor : MH12874
          TaqMan™ Advanced miRNA Assay : rno480942_mir
          TaqMan™ Advanced miRNA Assay : mmu482614_mir
          TaqMan™ Advanced miRNA Assay : 478717_mir


          miRBase Alias:

          Human: hsa-miR-147b (v21)
          Mouse: mmu-miR-147 (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline