Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478855_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-374a-3p
          Stem-loop Accession Number: MI0000782
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 74287286 - 74287357 [-] on Build GRCh38
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Species: Human, Callithrix jacchus, Dasypus novemcinctus, Nomascus leucogenys, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478855_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000288, mir-374

          Mature miRNA Sequence:

          CUUAUCAGAUUGUAUUGUAAUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-374a-3p MIMAT0004688
          Callithrix jacchus cja-miR-374a MIMAT0039407
          Dasypus novemcinctus dno-miR-374a-3p MIMAT0047847
          Nomascus leucogenys nle-miR-374a MIMAT0049414
          Rhesus monkey mml-miR-374a-3p MIMAT0026859

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-374a
          Stem-loop Accession # MI0000782
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUAUAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location Chr. X - 74287286 - 74287357 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004688
          miRBase ID: hsa-miR-374a-3p
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: Chr. X - 74287286 - 74287357 [-] on Build GRCh38

          Callithrix jacchus

          Stem-loop ID cja-mir-374a
          Stem-loop Accession # MI0031926
          Stem-loop Sequence
          GGAAAGAAAUUUUACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUAUAUGCCUGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039407
          miRBase ID: cja-miR-374a
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-374a
          Stem-loop Accession # MI0039088
          Stem-loop Sequence
          UUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047847
          miRBase ID: dno-miR-374a-3p
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-374a
          Stem-loop Accession # MI0040155
          Stem-loop Sequence
          UUAUAAUACAACCUGAUAAGUGUUAUAGCACUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049414
          miRBase ID: nle-miR-374a
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-374a
          Stem-loop Accession # MI0007718
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026859
          miRBase ID: mml-miR-374a-3p
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304235_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12702
          Ambion® Pre-miR™ miRNA Precursor : PM12702
          TaqMan™ MicroRNA Assay : 002125
          mirVana® miRNA inhibitor : MH12702
          mirVana® miRNA mimic : MC12702
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human B Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline