Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12702
          Assay Name: hsa-miR-374a-3p
          miRBase Accession Number: MI0000782
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 74287286 - 74287357 [-] on Build GRCh38
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Species: Human, Callithrix jacchus, Dasypus novemcinctus, Nomascus leucogenys, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12702
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000288, mir-374

          Mature miRNA Sequence:

          CUUAUCAGAUUGUAUUGUAAUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-374a-3p MIMAT0004688
          Callithrix jacchus cja-miR-374a MIMAT0039407
          Dasypus novemcinctus dno-miR-374a-3p MIMAT0047847
          Nomascus leucogenys nle-miR-374a MIMAT0049414
          Rhesus monkey mml-miR-374a-3p MIMAT0026859

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-374a
          Stem-loop Accession # MI0000782
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUAUAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location Chr. X - 74287286 - 74287357 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004688
          miRBase ID: hsa-miR-374a-3p
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: Chr. X - 74287286 - 74287357 [-] on Build GRCh38

          Callithrix jacchus

          Stem-loop ID cja-mir-374a
          Stem-loop Accession # MI0031926
          Stem-loop Sequence
          GGAAAGAAAUUUUACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUAUAUGCCUGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039407
          miRBase ID: cja-miR-374a
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-374a
          Stem-loop Accession # MI0039088
          Stem-loop Sequence
          UUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047847
          miRBase ID: dno-miR-374a-3p
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-374a
          Stem-loop Accession # MI0040155
          Stem-loop Sequence
          UUAUAAUACAACCUGAUAAGUGUUAUAGCACUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049414
          miRBase ID: nle-miR-374a
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-374a
          Stem-loop Accession # MI0007718
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026859
          miRBase ID: mml-miR-374a-3p
          Mature miRNA Sequence: CUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304235_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12702
          TaqMan™ MicroRNA Assay : 002125
          mirVana® miRNA inhibitor : MH12702
          mirVana® miRNA mimic : MC12702
          TaqMan™ Advanced miRNA Assay : 478855_mir


          miRBase Alias:

          Human: hsa-miR-374a* (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline