Search
Search

| Assay Name: | hsa-miR-374a-3p |
|---|---|
| miRBase Accession Number: | MI0000782 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUAUCAGAUUGUAUUGUAAUU |
| Species: | Human, Callithrix jacchus, Dasypus novemcinctus, Nomascus leucogenys, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | CUUAUCAGAUUGUAUUGUAAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-374a-3p | MIMAT0004688 |
| Callithrix jacchus | cja-miR-374a | MIMAT0039407 |
| Dasypus novemcinctus | dno-miR-374a-3p | MIMAT0047847 |
| Nomascus leucogenys | nle-miR-374a | MIMAT0049414 |
| Rhesus monkey | mml-miR-374a-3p | MIMAT0026859 |