Search
Search

| Assay Name: | hsa-miR-665 |
|---|---|
| Stem-loop Accession Number: | MI0005563 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCAGGAGGCUGAGGCCCCU |
| Species: | Human, Pan troglodytes, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000404, mir-665 |
Mature miRNA Sequence: | ACCAGGAGGCUGAGGCCCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-665 | MIMAT0004952 |
| Pan troglodytes | ptr-miR-665 | MIMAT0008312 |
| Rhesus monkey | mml-miR-665 | MIMAT0012786 |