Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002681
          Assay Name: hsa-miR-665
          miRBase Accession Number: MI0005563
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100875033 - 100875104 [+] on Build GRCh38
          Mature miRNA Sequence: ACCAGGAGGCUGAGGCCCCU
          Species: Human, Pan troglodytes, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002681
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000404, mir-665

          Mature miRNA Sequence:

          ACCAGGAGGCUGAGGCCCCU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-665 MIMAT0004952
          Pan troglodytes ptr-miR-665 MIMAT0008312
          Rhesus monkey mml-miR-665 MIMAT0012786

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-665
          Stem-loop Accession # MI0005563
          Stem-loop Sequence
          UCUCCUCGAGGGGUCUCUGCCUCUACCCAGGACUCUUUCAUGACCAGGAGGCUGAGGCCCCUCACAGGCGGC
          Chromosome Location Chr. 14 - 100875033 - 100875104 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004952
          miRBase ID: hsa-miR-665
          Mature miRNA Sequence: ACCAGGAGGCUGAGGCCCCU
          Chromosome Location: Chr. 14 - 100875033 - 100875104 [+] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-665
          Stem-loop Accession # MI0008849
          Stem-loop Sequence
          CUCCUGGAGGGGUCUCCGCCUCUACCCAGGACUCUUUCAUGACCAGGAGGCUGAGGCCCCUCACAGGCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008312
          miRBase ID: ptr-miR-665
          Mature miRNA Sequence: ACCAGGAGGCUGAGGCCCCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-665
          Stem-loop Accession # MI0012543
          Stem-loop Sequence
          UCUCCUCGAGGGGUCUCUGCCUCUACCCAGGACUCUUUCAUGACCAGGAGGCUGAGGCCCCUCACAGGCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012786
          miRBase ID: mml-miR-665
          Mature miRNA Sequence: ACCAGGAGGCUGAGGCCCCU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03293725_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12530
          Ambion® Pre-miR™ miRNA Precursor : PM12530
          mirVana® miRNA inhibitor : MH12530
          mirVana® miRNA mimic : MC12530
          TaqMan™ Advanced miRNA Assay : 479150_mir
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          FGMIR : TaqManTM Human MicroRNA Array B v3
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0