Search
Search

| Assay Name: | hsa-miR-28-5p |
|---|---|
| miRBase Accession Number: | MI0000086 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGAG |
| Species: | Human, Mouse, Rat, Ateles geoffroyi, Bovine, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Lagothrix lagotricha, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Saguinus labiatus, Tupaia chinensis |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-28-5p | MIMAT0000085 |
| Mouse | mmu-miR-28a-5p | MIMAT0000653 |
| Rat | rno-miR-28-5p | MIMAT0000800 |
| Ateles geoffroyi | age-miR-28 | MIMAT0002351 |
| Bovine | bta-miR-28 | MIMAT0009272 |
| Dasypus novemcinctus | dno-miR-28-5p | MIMAT0047547 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-28 | MIMAT0002354 |
| Guinea pig | cpo-miR-28-5p | MIMAT0046894 |
| Horse | eca-miR-28-5p | MIMAT0013092 |
| Lagothrix lagotricha | lla-miR-28 | MIMAT0002358 |
| Macaca nemestrina | mne-miR-28 | MIMAT0002356 |
| Pan paniscus | ppa-miR-28 | MIMAT0002359 |
| Pan troglodytes | ptr-miR-28 | MIMAT0002353 |
| Pig | ssc-miR-28-5p | MIMAT0002136 |
| Pongo pygmaeus | ppy-miR-28 | MIMAT0002355 |
| Rabbit | ocu-miR-28-5p | MIMAT0048141 |
| Rhesus monkey | mml-miR-28-5p | MIMAT0002352 |
| Saguinus labiatus | sla-miR-28 | MIMAT0002357 |
| Tupaia chinensis | tch-miR-28-5p | MIMAT0036565 |