Search
Search

| Assay Name: | hsa-miR-28 |
|---|---|
| miRBase Accession Number: | MI0000086 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGAG |
| Species: | Human, Mouse, Rat, Ateles geoffroyi, Bovine, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Lagothrix lagotricha, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Saguinus labiatus, Tupaia chinensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGAG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-28-5p | MIMAT0000085 | |
| Mouse | mmu-miR-28a-5p | MIMAT0000653 | |
| Rat | rno-miR-28-5p | MIMAT0000800 | |
| Ateles geoffroyi | age-miR-28 | MIMAT0002351 | |
| Bovine | bta-miR-28 | MIMAT0009272 | |
| Dasypus novemcinctus | dno-miR-28-5p | MIMAT0047547 | |
| View More Rows | |||
| Gorilla gorilla | ggo-miR-28 | MIMAT0002354 | |
| Guinea pig | cpo-miR-28-5p | MIMAT0046894 | |
| Horse | eca-miR-28-5p | MIMAT0013092 | |
| Lagothrix lagotricha | lla-miR-28 | MIMAT0002358 | |
| Macaca nemestrina | mne-miR-28 | MIMAT0002356 | |
| Pan paniscus | ppa-miR-28 | MIMAT0002359 | |
| Pan troglodytes | ptr-miR-28 | MIMAT0002353 | |
| Pig | ssc-miR-28-5p | MIMAT0002136 | |
| Pongo pygmaeus | ppy-miR-28 | MIMAT0002355 | |
| Rabbit | ocu-miR-28-5p | MIMAT0048141 | |
| Rhesus monkey | mml-miR-28-5p | MIMAT0002352 | |
| Saguinus labiatus | sla-miR-28 | MIMAT0002357 | |
| Tupaia chinensis | tch-miR-28-5p | MIMAT0036565 | |