Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm03307076_pri
Assay Name:
mmu-mir-28a
Stem-loop ID:
mmu-mir-28a
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.16: 24827855 - 24827940 [+] on Build GRCm38
Stem-loop Location:
Chr.16: 24827855 - 24827940 [+] on Build GRCm38
Mature miRNA ID:
mmu-miR-28a-5p
mmu-miR-28a-3p
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm03307076_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
69
Chromosome Location:
Chr. 16 - 24828255 - 24828255 [-] on 24828255 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-28a-5p
MIMAT0000653
Mouse
mmu-miR-28a-3p
MIMAT0004661
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-28a
Stem-loop Accession #
MI0000690
Stem-loop Sequence
GGUCCCUACCUUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGAUUCUCCCACUAGAUUGUGAGCUGCUGGAGGGCAGGCACU
Chromosome Location
Chr. 16 - 24827855 - 24827940 [+] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0000653
miRBase ID: mmu-miR-28a-5p
Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
Chromosome Location: Chr. 16 - 24827855 - 24827940 [+] on Build GRCm38
miRBase Accession #: MIMAT0004661
miRBase ID: mmu-miR-28a-3p
Mature miRNA Sequence: CACUAGAUUGUGAGCUGCUGGA
Chromosome Location: Chr. 16 - 24827855 - 24827940 [+] on Build GRCm38
Gene Family ID
MIPF0000057,
mir-28
Distance to Amplicon
280 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10346
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10346
TaqMan
™
MicroRNA Assay :
000411
mir
Vana
®
miRNA inhibitor :
MH10346
mir
Vana
®
miRNA mimic :
MC10346
TaqMan
™
Advanced miRNA Assay :
478000_mir
TaqMan
™
Advanced miRNA Assay :
mmu482619_mir
TaqMan
™
Advanced miRNA Assay :
rno481018_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM12298
Ambion
®
Pre-miR
™
miRNA Precursor :
PM12298
TaqMan
™
MicroRNA Assay :
002545
mir
Vana
®
miRNA inhibitor :
MH12298
mir
Vana
®
miRNA mimic :
MC12298
TaqMan
™
Advanced miRNA Assay :
mmu481665_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline