Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481018_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-28-5p
          Stem-loop Accession Number: MI0000086
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 188688781 - 188688866 [+] on Build GRCh38
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Species: Human, Mouse, Rat, Ateles geoffroyi, Bovine, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Lagothrix lagotricha, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Saguinus labiatus, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481018_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000057, mir-28

          Mature miRNA Sequence:

          AAGGAGCUCACAGUCUAUUGAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-28-5p MIMAT0000085
          Mouse mmu-miR-28a-5p MIMAT0000653
          Rat rno-miR-28-5p MIMAT0000800
          Ateles geoffroyi age-miR-28 MIMAT0002351
          Bovine bta-miR-28 MIMAT0009272
          Dasypus novemcinctus dno-miR-28-5p MIMAT0047547
          View More Rows
          Gorilla gorilla ggo-miR-28 MIMAT0002354
          Guinea pig cpo-miR-28-5p MIMAT0046894
          Horse eca-miR-28-5p MIMAT0013092
          Lagothrix lagotricha lla-miR-28 MIMAT0002358
          Macaca nemestrina mne-miR-28 MIMAT0002356
          Pan paniscus ppa-miR-28 MIMAT0002359
          Pan troglodytes ptr-miR-28 MIMAT0002353
          Pig ssc-miR-28-5p MIMAT0002136
          Pongo pygmaeus ppy-miR-28 MIMAT0002355
          Rabbit ocu-miR-28-5p MIMAT0048141
          Rhesus monkey mml-miR-28-5p MIMAT0002352
          Saguinus labiatus sla-miR-28 MIMAT0002357
          Tupaia chinensis tch-miR-28-5p MIMAT0036565

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-28
          Stem-loop Accession # MI0000086
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location Chr. 3 - 188688781 - 188688866 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000085
          miRBase ID: hsa-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: Chr. 3 - 188688781 - 188688866 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-28a
          Stem-loop Accession # MI0000690
          Stem-loop Sequence
          GGUCCCUACCUUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGAUUCUCCCACUAGAUUGUGAGCUGCUGGAGGGCAGGCACU
          Chromosome Location Chr. 16 - 24827855 - 24827940 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000653
          miRBase ID: mmu-miR-28a-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: Chr. 16 - 24827855 - 24827940 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-28
          Stem-loop Accession # MI0000861
          Stem-loop Sequence
          GGUCCCUACCCGCAAGGAGCUCACAGUCUAUUGAGUUCCUUUUCUGAUUCUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location Chr. 11 - 79380523 - 79380608 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000800
          miRBase ID: rno-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: Chr. 11 - 79380523 - 79380608 [-] on Build Rnor_6.0

          Ateles geoffroyi

          Stem-loop ID age-mir-28
          Stem-loop Accession # MI0002648
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002351
          miRBase ID: age-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-28
          Stem-loop Accession # MI0009785
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGAGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009272
          miRBase ID: bta-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-28
          Stem-loop Accession # MI0038929
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUUCCUUUUUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047547
          miRBase ID: dno-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-28
          Stem-loop Accession # MI0002651
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002354
          miRBase ID: ggo-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-28
          Stem-loop Accession # MI0038585
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUACUUUUAAGACUUUCCCACUAGAUUGGGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046894
          miRBase ID: cpo-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-28
          Stem-loop Accession # MI0012840
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013092
          miRBase ID: eca-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-28
          Stem-loop Accession # MI0002655
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002358
          miRBase ID: lla-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-28
          Stem-loop Accession # MI0002653
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUUCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002356
          miRBase ID: mne-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-28
          Stem-loop Accession # MI0002656
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002359
          miRBase ID: ppa-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-28
          Stem-loop Accession # MI0002650
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002353
          miRBase ID: ptr-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-28
          Stem-loop Accession # MI0002430
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGUCUUCCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002136
          miRBase ID: ssc-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-28
          Stem-loop Accession # MI0002652
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGAUUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002355
          miRBase ID: ppy-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-28
          Stem-loop Accession # MI0039242
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUGCCAUUCUGACUUCCCCACUAGAUUAUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048141
          miRBase ID: ocu-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-28
          Stem-loop Accession # MI0002649
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUUCCUUUGUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002352
          miRBase ID: mml-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-28
          Stem-loop Accession # MI0002654
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUAACUUUCCCACUAGAUUGUAAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002357
          miRBase ID: sla-miR-28
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-28
          Stem-loop Accession # MI0031226
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUGCCUUCCUGACUUUCUCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036565
          miRBase ID: tch-miR-28-5p
          Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465014_pri
          TaqMan™ Pri-miRNA Assay : Mm03307076_pri
          TaqMan™ Pri-miRNA Assay : Hs03302668_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10346
          Ambion® Pre-miR™ miRNA Precursor : PM10346
          TaqMan™ MicroRNA Assay : 000411
          mirVana® miRNA inhibitor : MH10346
          mirVana® miRNA mimic : MC10346
          TaqMan™ Advanced miRNA Assay : 478000_mir
          TaqMan™ Advanced miRNA Assay : mmu482619_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline