Search
Search

| Assay Name: | hsa-miR-151b |
|---|---|
| miRBase Accession Number: | MI0003772 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCGAGGAGCUCACAGUCU |
| Species: | Human |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | UCGAGGAGCUCACAGUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-151b | MIMAT0010214 |