Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03306902_pri
          Assay Name:
          mmu-mir-129-2
          Stem-loop ID:
          mmu-mir-129-2 mmu-mir-129b
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.2: 94241364 - 94241453 [-] on Build GRCm38
          Stem-loop Location:
          Chr.2: 94241364 - 94241453 [-] on Build GRCm38 Chr.2: 94241378 - 94241442 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-129-2-3p mmu-miR-129-5p mmu-miR-129b-5p mmu-miR-129b-3p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03306902_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          72

          Chromosome Location:

          Chr. 2 - 94241666 - 94241666 [+] on 94241666 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-129-2-3p MIMAT0000544
          Mouse mmu-miR-129-5p MIMAT0000209
          Mouse mmu-miR-129b-5p MIMAT0029862
          Mouse mmu-miR-129b-3p MIMAT0029863

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-129-2
          Stem-loop Accession # MI0000585
          Stem-loop Sequence
          UGCCUUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCG
          Chromosome Location Chr. 2 - 94241364 - 94241453 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000544
          miRBase ID: mmu-miR-129-2-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: Chr. 2 - 94241364 - 94241453 [-] on Build GRCm38

          miRBase Accession #: MIMAT0000209
          miRBase ID: mmu-miR-129-5p
          Mature miRNA Sequence: CUUUUUGCGGUCUGGGCUUGC
          Chromosome Location: Chr. 2 - 94241364 - 94241453 [-] on Build GRCm38

          Gene Family ID MIPF0000073, mir-129
          Distance to Amplicon 171 bases
          Stem-loop ID mmu-mir-129b
          Stem-loop Accession # MI0025019
          Stem-loop Sequence
          GCUUUUUGGGGUAAGGGCUUCCGGCUAUUGAGUUAUGUACAGCAAGCCCAGACCGCAAAAAGAUU
          Chromosome Location Chr. 2 - 94241378 - 94241442 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0029862
          miRBase ID: mmu-miR-129b-5p
          Mature miRNA Sequence: GCUUUUUGGGGUAAGGGCUUCC
          Chromosome Location: Chr. 2 - 94241378 - 94241442 [+] on Build GRCm38

          miRBase Accession #: MIMAT0029863
          miRBase ID: mmu-miR-129b-3p
          Mature miRNA Sequence: CAAGCCCAGACCGCAAAAAGAUU
          Chromosome Location: Chr. 2 - 94241378 - 94241442 [+] on Build GRCm38

          Gene Family ID MIPF0000073, mir-129
          Distance to Amplicon 182 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM28874
          Ambion® Pre-miR™ miRNA Precursor : PM28874
          TaqMan™ MicroRNA Assay : 467335_mat
          mirVana® miRNA inhibitor : MH28874
          mirVana® miRNA mimic : MC28874
          TaqMan™ Advanced miRNA Assay : mmu481535_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM26777
          Ambion® Pre-miR™ miRNA Precursor : PM26777
          TaqMan™ MicroRNA Assay : 466730_mat
          mirVana® miRNA inhibitor : MH26777
          mirVana® miRNA mimic : MC26777
          TaqMan™ Advanced miRNA Assay : mmu481534_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10076
          Ambion® Pre-miR™ miRNA Precursor : PM10076
          TaqMan™ MicroRNA Assay : 001184
          mirVana® miRNA inhibitor : MH10076
          mirVana® miRNA mimic : MC10076
          TaqMan™ Advanced miRNA Assay : rno481450_mir
          TaqMan™ Advanced miRNA Assay : mmu478544_mir
          TaqMan™ Advanced miRNA Assay : 478544_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10195
          Ambion® Pre-miR™ miRNA Precursor : PM10195
          TaqMan™ MicroRNA Assay : 000590
          mirVana® miRNA inhibitor : MH10195
          mirVana® miRNA mimic : MC10195
          TaqMan™ Advanced miRNA Assay : 477896_mir
          TaqMan™ Advanced miRNA Assay : rno480913_mir
          TaqMan™ Advanced miRNA Assay : mmu480913_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline