Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10076
          Assay Name: hsa-miR-129-2-3p
          miRBase Accession Number: MI0000473
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 43581394 - 43581483 [+] on Build GRCh38
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Bovine, Chicken, Dasypus novemcinctus, Eptesicus fuscus, Gadus morhua, Goat, Guinea pig, Horse, Ophiophagus hannah, Petromyzon marinus, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Salmo salar, Taeniopygia guttata, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC10076

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000073, mir-129

          Mature miRNA Sequence:

          AAGCCCUUACCCCAAAAAGCAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-129-2-3p MIMAT0004605
          Mouse mmu-miR-129-2-3p MIMAT0000544
          Rat rno-miR-129-2-3p MIMAT0000601
          Alligator mississippiensis ami-miR-129b-3p MIMAT0038139
          Bovine bta-miR-129-3p MIMAT0009222
          Chicken gga-miR-129-1-3p MIMAT0050057
          View More Rows
          Dasypus novemcinctus dno-miR-129-2-3p MIMAT0047626
          Eptesicus fuscus efu-miR-129a MIMAT0035001
          Gadus morhua gmo-miR-129-3p MIMAT0044075
          Goat chi-miR-129-3p MIMAT0035937
          Guinea pig cpo-miR-129-2-3p MIMAT0046979
          Horse eca-miR-129b-3p MIMAT0013046
          Ophiophagus hannah oha-miR-129b-3p MIMAT0036684
          Ophiophagus hannah oha-miR-129b MIMAT0036682
          Petromyzon marinus pma-miR-129a-3p MIMAT0019451
          Pig ssc-miR-129a-3p MIMAT0013959
          Pongo pygmaeus ppy-miR-129-2-3p MIMAT0016972
          Pteropus alecto pal-miR-129a-3p MIMAT0039855
          Python bivittatus pbv-miR-129b-3p MIMAT0038925
          Rabbit ocu-miR-129-2-3p MIMAT0048216
          Salmo salar ssa-miR-129-3p MIMAT0032319
          Taeniopygia guttata tgu-miR-129-3p MIMAT0014499
          Zebrafish dre-miR-129-3p MIMAT0003158

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-129-2
          Stem-loop Accession # MI0000473
          Stem-loop Sequence
          UGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUUGCGGAGGGCG
          Chromosome Location Chr. 11 - 43581394 - 43581483 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004605
          miRBase ID: hsa-miR-129-2-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: Chr. 11 - 43581394 - 43581483 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-129-2
          Stem-loop Accession # MI0000585
          Stem-loop Sequence
          UGCCUUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCG
          Chromosome Location Chr. 2 - 94241364 - 94241453 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000544
          miRBase ID: mmu-miR-129-2-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: Chr. 2 - 94241364 - 94241453 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-129-2
          Stem-loop Accession # MI0000637
          Stem-loop Sequence
          AGACUGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCGCGC
          Chromosome Location Chr. 3 - 83137297 - 83137393 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000601
          miRBase ID: rno-miR-129-2-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: Chr. 3 - 83137297 - 83137393 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-129b
          Stem-loop Accession # MI0029728
          Stem-loop Sequence
          CUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUACCUAGCCGGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038139
          miRBase ID: ami-miR-129b-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-129-2
          Stem-loop Accession # MI0009729
          Stem-loop Sequence
          CUGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009222
          miRBase ID: bta-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-129-1
          Stem-loop Accession # MI0040749
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUACAUAACUACCUACCCGGGAAGCCCUUACCCCAAAAAGCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0050057
          miRBase ID: gga-miR-129-1-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-129-2
          Stem-loop Accession # MI0038973
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047626
          miRBase ID: dno-miR-129-2-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-129a
          Stem-loop Accession # MI0028686
          Stem-loop Sequence
          CCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035001
          miRBase ID: efu-miR-129a
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-129-1
          Stem-loop Accession # MI0036121
          Stem-loop Sequence
          UCUUUUUGCGGUCUGGGCUUGCUGUUCAUAACUAAUAAGCUGGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044075
          miRBase ID: gmo-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Stem-loop ID gmo-mir-129-2
          Stem-loop Accession # MI0036166
          Stem-loop Sequence
          UCUUUUUGCGGUCUGGGCUUGCUGUUCCCACAGCCUUAGCCAGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044075
          miRBase ID: gmo-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Stem-loop ID gmo-mir-129-3
          Stem-loop Accession # MI0036019
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUGAUCGUGAUCUGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044075
          miRBase ID: gmo-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Stem-loop ID gmo-mir-129-4
          Stem-loop Accession # MI0036109
          Stem-loop Sequence
          ACUUUUUGCGGUCUGGGCUUGCUGUCCAAUCACUAUCCAGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044075
          miRBase ID: gmo-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-129
          Stem-loop Accession # MI0030612
          Stem-loop Sequence
          AAGCCGGCAGACUCUGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035937
          miRBase ID: chi-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-129-2
          Stem-loop Accession # MI0038632
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUACAUUACUCAUAGCCGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046979
          miRBase ID: cpo-miR-129-2-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-129b
          Stem-loop Accession # MI0012789
          Stem-loop Sequence
          GCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013046
          miRBase ID: eca-miR-129b-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-129b-1
          Stem-loop Accession # MI0031333
          Stem-loop Sequence
          AUGACGGGUCUUCUCUGGGUCUUUUUGCGGUCUGGGCUUUCUGGGUCUGUCCAACCCCGUAUCAGAAAGCCCUUACCCCAAAAAGCAUCUGGGGUUGUACAAUGGCAUCGCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036684
          miRBase ID: oha-miR-129b-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Stem-loop ID oha-mir-129b-2
          Stem-loop Accession # MI0031332
          Stem-loop Sequence
          ACUUUGUCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUAUAUAAGUACCUAGCCGGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCGCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036682
          miRBase ID: oha-miR-129b
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Petromyzon marinus

          Stem-loop ID pma-mir-129a
          Stem-loop Accession # MI0017062
          Stem-loop Sequence
          UGUCUCUCGCGAAUCUUUUUGCGGUUUGGGCUUGCUGUUCAUCUAAUAGCCGGCAAGCCCUUACCCCAAAAAGCAUUCGAGGGGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019451
          miRBase ID: pma-miR-129a-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-129a
          Stem-loop Accession # MI0013169
          Stem-loop Sequence
          CUUCGUGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013959
          miRBase ID: ssc-miR-129a-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-129-2
          Stem-loop Accession # MI0014821
          Stem-loop Sequence
          UGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUUGCGGAGGGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016972
          miRBase ID: ppy-miR-129-2-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-129a
          Stem-loop Accession # MI0032365
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039855
          miRBase ID: pal-miR-129a-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-129b
          Stem-loop Accession # MI0030222
          Stem-loop Sequence
          GUCUUCUCUGGGUCUUUUUGCGGUCUGGGCUUUCUGGGUCUGUCCAACCCCGUAUCAGAAAGCCCUUACCCCAAAAAGCAUCCGGGGUUGUACGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038925
          miRBase ID: pbv-miR-129b-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-129-2
          Stem-loop Accession # MI0039284
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048216
          miRBase ID: ocu-miR-129-2-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-129-1
          Stem-loop Accession # MI0026465
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUUCAUAAAUAUCAGUCUGGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032319
          miRBase ID: ssa-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-129-2
          Stem-loop Accession # MI0026466
          Stem-loop Sequence
          CUUUUUGCGGUCUGGGCUUGCUGUUAACACACCUCAACCCAGGAAGCCCUUACCCCAAAAAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032319
          miRBase ID: ssa-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-129
          Stem-loop Accession # MI0013688
          Stem-loop Sequence
          CUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUACCUACCCGGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014499
          miRBase ID: tgu-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-129-2
          Stem-loop Accession # MI0001982
          Stem-loop Sequence
          GUCUUUCACGAAUCUUUUUGCGGUCUGGGCUUGCUGUUCUCAACUAUCAAUGGGAAGCCCUUACCCCAAAAAGCAUUUGCGGAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003158
          miRBase ID: dre-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA

          Stem-loop ID dre-mir-129-4
          Stem-loop Accession # MI0039508
          Stem-loop Sequence
          GUCUUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUUCAUAAGUAUCUAGCUGGGAAGCCCUUACCCCAAAAAGCAUUUGCGGAAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003158
          miRBase ID: dre-miR-129-3p
          Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306902_pri
          TaqMan™ Pri-miRNA Assay : Hs03303241_pri
          TaqMan™ Pri-miRNA Assay : Rn03464502_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10076
          Ambion® Pre-miR™ miRNA Precursor : PM10076
          TaqMan™ MicroRNA Assay : 001184
          mirVana® miRNA inhibitor : MH10076
          TaqMan™ Advanced miRNA Assay : rno481450_mir
          TaqMan™ Advanced miRNA Assay : mmu478544_mir
          TaqMan™ Advanced miRNA Assay : 478544_mir


          miRBase Alias:

          Human: hsa-miR-129-3p (v17)
          Rat: rno-miR-129-2* (v18)
          Danio rerio: dre-miR-129* (v18)
          Phocea massiliensis: pma-miR-129a* (v18)
          Pongo pygmaeus: ppy-miR-129-2* (v18)
          Staphylococcus schleiferi: ssc-miR-129 (v16)
          Staphylococcus schleiferi: ssc-miR-129a (v20)
          Trabulsiella guamensis ATCC 49490: tgu-miR-129 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline