Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04272630_pri
          Assay Name:
          mmu-mir-467c
          Stem-loop ID:
          mmu-mir-466b-1 mmu-mir-466o mmu-mir-467c mmu-mir-669a-3 mmu-mir-669k
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.2: 10474219 - 10474300 [+] on Build GRCm38
          Stem-loop Location:
          Chr.2: 10474219 - 10474300 [+] on Build GRCm38 Chr.2: 10472540 - 10472623 [+] on Build GRCm38 Chr.2: 10473931 - 10474027 [+] on Build GRCm38 Chr.2: 10474433 - 10474541 [+] on Build GRCm38 Chr.2: 10475300 - 10475426 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-466b-3p mmu-miR-466b-5p mmu-miR-466o-5p mmu-miR-466o-3p mmu-miR-467c-3p mmu-miR-467c-5p mmu-miR-669a-5p mmu-miR-669a-3-3p mmu-miR-669k-3p mmu-miR-669k-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm04272630_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          165

          Chromosome Location:

          Chr. 2 - 10473712 - 10473712 [+] on 10473712 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-466b-3p MIMAT0004876
          Mouse mmu-miR-466b-5p MIMAT0004875
          Mouse mmu-miR-466o-5p MIMAT0014885
          Mouse mmu-miR-466o-3p MIMAT0014886
          Mouse mmu-miR-467c-3p MIMAT0017275
          Mouse mmu-miR-467c-5p MIMAT0004885
          View More Rows
          Mouse mmu-miR-669a-5p MIMAT0003477
          Mouse mmu-miR-669a-3-3p MIMAT0017251
          Mouse mmu-miR-669k-3p MIMAT0005831
          Mouse mmu-miR-669k-5p MIMAT0017323

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-466b-1
          Stem-loop Accession # MI0005502
          Stem-loop Sequence
          UGUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAG
          Chromosome Location Chr. 2 - 10474219 - 10474300 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10474219 - 10474300 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004875
          miRBase ID: mmu-miR-466b-5p
          Mature miRNA Sequence: UGAUGUGUGUGUACAUGUACAU
          Chromosome Location: Chr. 2 - 10474219 - 10474300 [+] on Build GRCm38

          Gene Family ID MIPF0000208, mir-466
          Distance to Amplicon 453 bases
          Stem-loop ID mmu-mir-466o
          Stem-loop Accession # MI0014052
          Stem-loop Sequence
          UGUGUGUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAUACAUGCACACAUAAGACACAUAUGAGCACA
          Chromosome Location Chr. 2 - 10472540 - 10472623 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0014885
          miRBase ID: mmu-miR-466o-5p
          Mature miRNA Sequence: UGAUGUGUGUGUACAUGUACAU
          Chromosome Location: Chr. 2 - 10472540 - 10472623 [+] on Build GRCm38

          miRBase Accession #: MIMAT0014886
          miRBase ID: mmu-miR-466o-3p
          Mature miRNA Sequence: UACAUACAUGCACACAUAAGAC
          Chromosome Location: Chr. 2 - 10472540 - 10472623 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 979 bases
          Stem-loop ID mmu-mir-467c
          Stem-loop Accession # MI0005512
          Stem-loop Sequence
          CCUUUGUGCAUAAGUGCGUGCAUGUAUAUGUGUGUAUAUUUUAUGCAUAUACAUACACACACCUAUACACACAUGCACACAGACAUGCGAGAAUGGC
          Chromosome Location Chr. 2 - 10473931 - 10474027 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017275
          miRBase ID: mmu-miR-467c-3p
          Mature miRNA Sequence: AUAUACAUACACACACCUAUAC
          Chromosome Location: Chr. 2 - 10473931 - 10474027 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004885
          miRBase ID: mmu-miR-467c-5p
          Mature miRNA Sequence: UAAGUGCGUGCAUGUAUAUGUG
          Chromosome Location: Chr. 2 - 10473931 - 10474027 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 165 bases
          Stem-loop ID mmu-mir-669a-3
          Stem-loop Accession # MI0004668
          Stem-loop Sequence
          UUCCUCCAUGUAUGUGCAUGUGUGUAUAGUUGUGUGUGCAUGUUCAUGUCUAUAUUUGAAUAUACAUAACAUACACACACAUGUAUAAACGCAAGCACACAUACACAGA
          Chromosome Location Chr. 2 - 10474433 - 10474541 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003477
          miRBase ID: mmu-miR-669a-5p
          Mature miRNA Sequence: AGUUGUGUGUGCAUGUUCAUGUCU
          Chromosome Location: Chr. 2 - 10474433 - 10474541 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017251
          miRBase ID: mmu-miR-669a-3-3p
          Mature miRNA Sequence: ACAUAACAUACACACACAUGUAU
          Chromosome Location: Chr. 2 - 10474433 - 10474541 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 667 bases
          Stem-loop ID mmu-mir-669k
          Stem-loop Accession # MI0006279
          Stem-loop Sequence
          UCUUAUGCCCACCAAAAAUGUGCAUGUGUGUAUAGUUGUGUGCAUAUUCACUUCUCUAUAUGAAUAUGCAUAUACACGCAUGCAAACACAUUCAUAGGAAUGGCCAGAGCUAUAUCUCAGUUAGGAG
          Chromosome Location Chr. 2 - 10475300 - 10475426 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0005831
          miRBase ID: mmu-miR-669k-3p
          Mature miRNA Sequence: UAUGCAUAUACACGCAUGCAA
          Chromosome Location: Chr. 2 - 10475300 - 10475426 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017323
          miRBase ID: mmu-miR-669k-5p
          Mature miRNA Sequence: UGUGCAUGUGUGUAUAGUUGUGUGC
          Chromosome Location: Chr. 2 - 10475300 - 10475426 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 1534 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM19674
          Ambion® Pre-miR™ miRNA Precursor : PM19674
          TaqMan™ MicroRNA Assay : 462897_mat
          mirVana® miRNA inhibitor : MH19674
          mirVana® miRNA mimic : MC19674
          Ambion® Anti-miR™ miRNA Inhibitor : AM20178
          Ambion® Pre-miR™ miRNA Precursor : PM20178
          TaqMan™ MicroRNA Assay : 464331_mat
          mirVana® miRNA inhibitor : MH20178
          mirVana® miRNA mimic : MC20178
          Ambion® Anti-miR™ miRNA Inhibitor : AM12358
          Ambion® Pre-miR™ miRNA Precursor : PM12358
          TaqMan™ MicroRNA Assay : 002517
          mirVana® miRNA inhibitor : MH12358
          mirVana® miRNA mimic : MC12358
          Ambion® Anti-miR™ miRNA Inhibitor : AM20509
          Ambion® Pre-miR™ miRNA Precursor : PM20509
          mirVana® miRNA inhibitor : MH20509
          mirVana® miRNA mimic : MC20509
          TaqMan™ Advanced miRNA Assay : mmu482917_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19878
          Ambion® Pre-miR™ miRNA Precursor : PM19878
          TaqMan™ MicroRNA Assay : 462148_mat
          mirVana® miRNA inhibitor : MH19878
          mirVana® miRNA mimic : MC19878
          Ambion® Anti-miR™ miRNA Inhibitor : AM20138
          Ambion® Pre-miR™ miRNA Precursor : PM20138
          TaqMan™ MicroRNA Assay : 462339_mat
          mirVana® miRNA inhibitor : MH20138
          mirVana® miRNA mimic : MC20138
          TaqMan™ Advanced miRNA Assay : mmu482771_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13506
          Ambion® Pre-miR™ miRNA Precursor : PM13506
          TaqMan™ MicroRNA Assay : 002900
          mirVana® miRNA inhibitor : MH13506
          mirVana® miRNA mimic : MC13506
          Ambion® Anti-miR™ miRNA Inhibitor : AM19867
          Ambion® Pre-miR™ miRNA Precursor : PM19867
          TaqMan™ MicroRNA Assay : 462521_mat
          mirVana® miRNA inhibitor : MH19867
          mirVana® miRNA mimic : MC19867
          TaqMan™ Advanced miRNA Assay : mmu482045_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19359
          Ambion® Pre-miR™ miRNA Precursor : PM19359
          TaqMan™ MicroRNA Assay : 464896_mat
          mirVana® miRNA inhibitor : MH19359
          mirVana® miRNA mimic : MC19359
          TaqMan™ Advanced miRNA Assay : mmu482642_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline