Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu482642_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-466b-3p
          Stem-loop Accession Number: MI0005502
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 10474219 - 10474300 [+] on Build GRCm38
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Species: Mouse
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu482642_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000208, mir-466

          Mature miRNA Sequence:

          AUACAUACACGCACACAUAAGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-466b-3p MIMAT0004876
          Mouse mmu-miR-466c-3p MIMAT0004878
          Mouse mmu-miR-466p-3p MIMAT0014892

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-466b-1
          Stem-loop Accession # MI0005502
          Stem-loop Sequence
          UGUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAG
          Chromosome Location Chr. 2 - 10474219 - 10474300 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466b-2
          Stem-loop Accession # MI0005503
          Stem-loop Sequence
          UGUAUGUGUUGAUGUGUGUGUACAUGUACAUGUCUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAG
          Chromosome Location Chr. 2 - 10498685 - 10498766 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466b-3
          Stem-loop Accession # MI0005504
          Stem-loop Sequence
          UAUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAAAUACAUACACGCACACAUAAGACACAUAUGA
          Chromosome Location Chr. 2 - 10503565 - 10503645 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466b-4
          Stem-loop Accession # MI0014060
          Stem-loop Sequence
          UGUGUGUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGCACA
          Chromosome Location Chr. 2 - 10486423 - 10486512 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466b-5
          Stem-loop Accession # MI0014063
          Stem-loop Sequence
          UGUGUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGCACA
          Chromosome Location Chr. 2 - 10488887 - 10488974 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466b-6
          Stem-loop Accession # MI0014067
          Stem-loop Sequence
          UGUGUGUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGCACA
          Chromosome Location Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466b-7
          Stem-loop Accession # MI0014070
          Stem-loop Sequence
          UGUGUAUGUGUUGAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGCACA
          Chromosome Location Chr. 2 - 10496268 - 10496355 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466b-8
          Stem-loop Accession # MI0014112
          Stem-loop Sequence
          GUGUGUAUGUGUUGAUGUGUGUGUACAUGUACAUAUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGC
          Chromosome Location Chr. 2 - 10476628 - 10476713 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10493798 - 10493887 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466c-1
          Stem-loop Accession # MI0005505
          Stem-loop Sequence
          GUAUAUGUGUUGAUGUGUGUGUGCAUGUACAUAUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGC
          Chromosome Location Chr. 2 - 10481534 - 10481617 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004878
          miRBase ID: mmu-miR-466c-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10481534 - 10481617 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466c-2
          Stem-loop Accession # MI0014057
          Stem-loop Sequence
          UGUGUAUAUGUGUUGAUGUGUGUGUGCAUGUACAUAUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGCACA
          Chromosome Location Chr. 2 - 10483966 - 10484055 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004878
          miRBase ID: mmu-miR-466c-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10481534 - 10481617 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466c-3
          Stem-loop Accession # MI0031753
          Stem-loop Sequence
          UGUGUAUAUGUGUUGAUGUGUGUGUGCAUGUACAUAUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGCACA
          Chromosome Location Chr. 2 - 10491336 - 10491425 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004878
          miRBase ID: mmu-miR-466c-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10481534 - 10481617 [+] on Build GRCm38

          Stem-loop ID mmu-mir-466p
          Stem-loop Accession # MI0014078
          Stem-loop Sequence
          UGUGUAUAUGUGUUUAUGUGUGUGUACAUGUACAUGUGUGAAUAUGAUGAUAUAUACAUACACGCACACAUAAGACACAUAUGAGCACA
          Chromosome Location Chr. 2 - 10506006 - 10506094 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0014892
          miRBase ID: mmu-miR-466p-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10506006 - 10506094 [+] on Build GRCm38


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm04272635_pri
          TaqMan™ Pri-miRNA Assay : Mm04272633_pri
          TaqMan™ Pri-miRNA Assay : Mm03308706_pri
          TaqMan™ Pri-miRNA Assay : Mm04272630_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19359
          Ambion® Pre-miR™ miRNA Precursor : PM19359
          TaqMan™ MicroRNA Assay : 464896_mat
          mirVana® miRNA inhibitor : MH19359
          mirVana® miRNA mimic : MC19359

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline