Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04272633_pri
          Assay Name:
          mmu-mir-467a-2
          Stem-loop ID:
          mmu-mir-466b-8 mmu-mir-466c-1 mmu-mir-466e mmu-mir-467a-1 mmu-mir-467a-2 mmu-mir-467b mmu-mir-669a-1 mmu-mir-669a-4 mmu-mir-669a-5 mmu-mir-669g mmu-mir-669j
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.2: 10476628 - 10476713 [+] on Build GRCm38
          Stem-loop Location:
          Chr.2: 10476628 - 10476713 [+] on Build GRCm38 Chr.2: 10481534 - 10481617 [+] on Build GRCm38 Chr.2: 10479088 - 10479171 [+] on Build GRCm38 Chr.2: 10476346 - 10476418 [+] on Build GRCm38 Chr.2: 10478798 - 10478880 [+] on Build GRCm38 Chr.2: 10481248 - 10481320 [+] on Build GRCm38 Chr.2: 10476853 - 10476949 [+] on Build GRCm38 Chr.2: 10479315 - 10479401 [+] on Build GRCm38 Chr.2: 10481761 - 10481847 [+] on Build GRCm38 Chr.2: 10477150 - 10477272 [+] on Build GRCm38 Chr.2: 10477910 - 10478030 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-466b-3p mmu-miR-466b-5p mmu-miR-466c-3p mmu-miR-466c-5p mmu-miR-466e-3p mmu-miR-466e-5p mmu-miR-467a-5p mmu-miR-467a-3p mmu-miR-467b-5p mmu-miR-467b-3p mmu-miR-669a-5p mmu-miR-669a-3p mmu-miR-669g mmu-miR-669j
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm04272633_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          122

          Chromosome Location:

          Chr. 2 - 10478540 - 10478540 [+] on 10478540 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-466b-3p MIMAT0004876
          Mouse mmu-miR-466b-5p MIMAT0004875
          Mouse mmu-miR-466c-3p MIMAT0004878
          Mouse mmu-miR-466c-5p MIMAT0004877
          Mouse mmu-miR-466e-3p MIMAT0004880
          Mouse mmu-miR-466e-5p MIMAT0004879
          View More Rows
          Mouse mmu-miR-467a-5p MIMAT0003409
          Mouse mmu-miR-467a-3p MIMAT0002108
          Mouse mmu-miR-467b-5p MIMAT0005448
          Mouse mmu-miR-467b-3p MIMAT0003478
          Mouse mmu-miR-669a-5p MIMAT0003477
          Mouse mmu-miR-669a-3p MIMAT0017243
          Mouse mmu-miR-669g MIMAT0005832
          Mouse mmu-miR-669j MIMAT0005838

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-466b-8
          Stem-loop Accession # MI0014112
          Stem-loop Sequence
          GUGUGUAUGUGUUGAUGUGUGUGUACAUGUACAUAUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGC
          Chromosome Location Chr. 2 - 10476628 - 10476713 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004876
          miRBase ID: mmu-miR-466b-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10476628 - 10476713 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004875
          miRBase ID: mmu-miR-466b-5p
          Mature miRNA Sequence: UGAUGUGUGUGUACAUGUACAU
          Chromosome Location: Chr. 2 - 10476628 - 10476713 [+] on Build GRCm38

          Gene Family ID MIPF0000208, mir-466
          Distance to Amplicon 1759 bases
          Stem-loop ID mmu-mir-466c-1
          Stem-loop Accession # MI0005505
          Stem-loop Sequence
          GUAUAUGUGUUGAUGUGUGUGUGCAUGUACAUAUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGC
          Chromosome Location Chr. 2 - 10481534 - 10481617 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004878
          miRBase ID: mmu-miR-466c-3p
          Mature miRNA Sequence: AUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10481534 - 10481617 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004877
          miRBase ID: mmu-miR-466c-5p
          Mature miRNA Sequence: UGAUGUGUGUGUGCAUGUACAUAU
          Chromosome Location: Chr. 2 - 10481534 - 10481617 [+] on Build GRCm38

          Gene Family ID MIPF0000208, mir-466
          Distance to Amplicon 2941 bases
          Stem-loop ID mmu-mir-466e
          Stem-loop Accession # MI0005506
          Stem-loop Sequence
          GUGUAUGUGUUGAUGUGUGUGUACAUGUACAUAUGUGAAUAUGAUAUACAUAUACAUACACGCACACAUAAGACACAUAUGAGC
          Chromosome Location Chr. 2 - 10479088 - 10479171 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004880
          miRBase ID: mmu-miR-466e-3p
          Mature miRNA Sequence: UAUACAUACACGCACACAUAAGA
          Chromosome Location: Chr. 2 - 10479088 - 10479171 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004879
          miRBase ID: mmu-miR-466e-5p
          Mature miRNA Sequence: GAUGUGUGUGUACAUGUACAUA
          Chromosome Location: Chr. 2 - 10479088 - 10479171 [+] on Build GRCm38

          Gene Family ID MIPF0000208, mir-466
          Distance to Amplicon 495 bases
          Stem-loop ID mmu-mir-467a-1
          Stem-loop Accession # MI0002402
          Stem-loop Sequence
          CUGUGUGCGUAAGUGCCUGCAUGUAUAUGCGUGUAUAUUUUAUGCAUAUACAUACACACACCUACACACACAU
          Chromosome Location Chr. 2 - 10476346 - 10476418 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003409
          miRBase ID: mmu-miR-467a-5p
          Mature miRNA Sequence: UAAGUGCCUGCAUGUAUAUGCG
          Chromosome Location: Chr. 2 - 10476346 - 10476418 [+] on Build GRCm38

          miRBase Accession #: MIMAT0002108
          miRBase ID: mmu-miR-467a-3p
          Mature miRNA Sequence: CAUAUACAUACACACACCUACA
          Chromosome Location: Chr. 2 - 10476346 - 10476418 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 2054 bases
          Stem-loop ID mmu-mir-467a-2
          Stem-loop Accession # MI0014053
          Stem-loop Sequence
          UGUGCCUGUGUGCGUAAGUGCCUGCAUGUAUAUGCGUGUAUAUUUUAUGCAUAUACAUACACACACCUACACACACAUGCACA
          Chromosome Location Chr. 2 - 10478798 - 10478880 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003409
          miRBase ID: mmu-miR-467a-5p
          Mature miRNA Sequence: UAAGUGCCUGCAUGUAUAUGCG
          Chromosome Location: Chr. 2 - 10476346 - 10476418 [+] on Build GRCm38

          miRBase Accession #: MIMAT0002108
          miRBase ID: mmu-miR-467a-3p
          Mature miRNA Sequence: CAUAUACAUACACACACCUACA
          Chromosome Location: Chr. 2 - 10476346 - 10476418 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 205 bases
          Stem-loop ID mmu-mir-467b
          Stem-loop Accession # MI0004671
          Stem-loop Sequence
          CCGUGUGCGUAAGUGCCUGCAUGUAUAUGCGUGUAUAUUUUAUGCAUAUACAUACACACACCAACACACACAU
          Chromosome Location Chr. 2 - 10481248 - 10481320 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0005448
          miRBase ID: mmu-miR-467b-5p
          Mature miRNA Sequence: GUAAGUGCCUGCAUGUAUAUG
          Chromosome Location: Chr. 2 - 10481248 - 10481320 [+] on Build GRCm38

          miRBase Accession #: MIMAT0003478
          miRBase ID: mmu-miR-467b-3p
          Mature miRNA Sequence: AUAUACAUACACACACCAACAC
          Chromosome Location: Chr. 2 - 10481248 - 10481320 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 2655 bases
          Stem-loop ID mmu-mir-669a-1
          Stem-loop Accession # MI0004523
          Stem-loop Sequence
          UGUAUGUGCAUGUGUGUAUAGUUGUGUGUGCAUGUUCAUGUCUAUAUUUGAAUAUACAUAACAUACACACACACGUAUAAACGCAAGCACACAUACA
          Chromosome Location Chr. 2 - 10476853 - 10476949 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003477
          miRBase ID: mmu-miR-669a-5p
          Mature miRNA Sequence: AGUUGUGUGUGCAUGUUCAUGUCU
          Chromosome Location: Chr. 2 - 10476853 - 10476949 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017243
          miRBase ID: mmu-miR-669a-3p
          Mature miRNA Sequence: ACAUAACAUACACACACACGUAU
          Chromosome Location: Chr. 2 - 10476853 - 10476949 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 1523 bases
          Stem-loop ID mmu-mir-669a-4
          Stem-loop Accession # MI0014054
          Stem-loop Sequence
          UGUGCAUGUGUGUAUAGUUGUGUGUGCAUGUUCAUGUCUAUAUUUGAAUAUACAUAACAUACACACACACGUAUAAACGCAAGCACA
          Chromosome Location Chr. 2 - 10479315 - 10479401 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003477
          miRBase ID: mmu-miR-669a-5p
          Mature miRNA Sequence: AGUUGUGUGUGCAUGUUCAUGUCU
          Chromosome Location: Chr. 2 - 10476853 - 10476949 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017243
          miRBase ID: mmu-miR-669a-3p
          Mature miRNA Sequence: ACAUAACAUACACACACACGUAU
          Chromosome Location: Chr. 2 - 10476853 - 10476949 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 722 bases
          Stem-loop ID mmu-mir-669a-5
          Stem-loop Accession # MI0014055
          Stem-loop Sequence
          UGUGCAUGUGUGUAUAGUUGUGUGUGCAUGUUCAUGUCUAUAUUUGAAUAUACAUAACAUACACACACACGUAUAAACGCAAGCACA
          Chromosome Location Chr. 2 - 10481761 - 10481847 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003477
          miRBase ID: mmu-miR-669a-5p
          Mature miRNA Sequence: AGUUGUGUGUGCAUGUUCAUGUCU
          Chromosome Location: Chr. 2 - 10476853 - 10476949 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017243
          miRBase ID: mmu-miR-669a-3p
          Mature miRNA Sequence: ACAUAACAUACACACACACGUAU
          Chromosome Location: Chr. 2 - 10476853 - 10476949 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 3168 bases
          Stem-loop ID mmu-mir-669g
          Stem-loop Accession # MI0006280
          Stem-loop Sequence
          CAUGUGUGCUUACUUGUAUGUGCAUUGUAUGUGUUGACAUGAUAUACAUAACAUAAACACACGUGUAUCUGCAAGCACACACACACAGGAAAAAUUGCAUUCAUUUAUACGUUUGAAUGAUAU
          Chromosome Location Chr. 2 - 10477150 - 10477272 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0005832
          miRBase ID: mmu-miR-669g
          Mature miRNA Sequence: UGCAUUGUAUGUGUUGACAUGAU
          Chromosome Location: Chr. 2 - 10477150 - 10477272 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 1200 bases
          Stem-loop ID mmu-mir-669j
          Stem-loop Accession # MI0006286
          Stem-loop Sequence
          AGCAUGCGAUUCAUGUGAUGUCCACCAAGAAUGUGCAUGUGUGUAUAGUUGUGUACAUAUUCACUUUUGUAUAUGAAUAUGCAUAUACUCACAUGCAAACAUACUCCCACGAAUGGCCCGA
          Chromosome Location Chr. 2 - 10477910 - 10478030 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0005838
          miRBase ID: mmu-miR-669j
          Mature miRNA Sequence: UGCAUAUACUCACAUGCAAACA
          Chromosome Location: Chr. 2 - 10477910 - 10478030 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 442 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12639
          Ambion® Pre-miR™ miRNA Precursor : PM12639
          TaqMan™ MicroRNA Assay : 002587
          mirVana® miRNA inhibitor : MH12639
          mirVana® miRNA mimic : MC12639
          TaqMan™ Advanced miRNA Assay : mmu483234_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20509
          Ambion® Pre-miR™ miRNA Precursor : PM20509
          mirVana® miRNA inhibitor : MH20509
          mirVana® miRNA mimic : MC20509
          TaqMan™ Advanced miRNA Assay : mmu482917_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19687
          Ambion® Pre-miR™ miRNA Precursor : PM19687
          TaqMan™ MicroRNA Assay : 464179_mat
          mirVana® miRNA inhibitor : MH19687
          mirVana® miRNA mimic : MC19687
          TaqMan™ Advanced miRNA Assay : mmu482998_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19954
          Ambion® Pre-miR™ miRNA Precursor : PM19954
          TaqMan™ MicroRNA Assay : 463964_mat
          mirVana® miRNA inhibitor : MH19954
          mirVana® miRNA mimic : MC19954
          Ambion® Anti-miR™ miRNA Inhibitor : AM13657
          Ambion® Pre-miR™ miRNA Precursor : PM13657
          TaqMan™ MicroRNA Assay : 002813
          mirVana® miRNA inhibitor : MH13657
          mirVana® miRNA mimic : MC13657
          TaqMan™ Advanced miRNA Assay : mmu482043_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12989
          Ambion® Pre-miR™ miRNA Precursor : PM12989
          TaqMan™ MicroRNA Assay : 002586
          mirVana® miRNA inhibitor : MH12989
          mirVana® miRNA mimic : MC12989
          Ambion® Anti-miR™ miRNA Inhibitor : AM12464
          Ambion® Pre-miR™ miRNA Precursor : PM12464
          TaqMan™ MicroRNA Assay : 002718
          mirVana® miRNA inhibitor : MH12464
          mirVana® miRNA mimic : MC12464
          Ambion® Anti-miR™ miRNA Inhibitor : AM19359
          Ambion® Pre-miR™ miRNA Precursor : PM19359
          TaqMan™ MicroRNA Assay : 464896_mat
          mirVana® miRNA inhibitor : MH19359
          mirVana® miRNA mimic : MC19359
          TaqMan™ Advanced miRNA Assay : mmu482642_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19405
          Ambion® Pre-miR™ miRNA Precursor : PM19405
          TaqMan™ MicroRNA Assay : 463771_mat
          mirVana® miRNA inhibitor : MH19405
          mirVana® miRNA mimic : MC19405
          Ambion® Anti-miR™ miRNA Inhibitor : AM19674
          Ambion® Pre-miR™ miRNA Precursor : PM19674
          TaqMan™ MicroRNA Assay : 462897_mat
          mirVana® miRNA inhibitor : MH19674
          mirVana® miRNA mimic : MC19674
          Ambion® Anti-miR™ miRNA Inhibitor : AM13655
          Ambion® Pre-miR™ miRNA Precursor : PM13655
          TaqMan™ MicroRNA Assay : 002899
          mirVana® miRNA inhibitor : MH13655
          mirVana® miRNA mimic : MC13655
          TaqMan™ Advanced miRNA Assay : mmu481932_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM11605
          Ambion® Pre-miR™ miRNA Precursor : PM11605
          TaqMan™ MicroRNA Assay : 001671
          mirVana® miRNA inhibitor : MH11605
          mirVana® miRNA mimic : MC11605
          TaqMan™ Advanced miRNA Assay : mmu482773_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM11433
          Ambion® Pre-miR™ miRNA Precursor : PM11433
          TaqMan™ MicroRNA Assay : 001684
          mirVana® miRNA inhibitor : MH11433
          mirVana® miRNA mimic : MC11433

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline