Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11007
          Assay Name:
          miRBase Accession Number: MI0010335
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Species: Chicken, Chrysemys picta, Dog, Gorilla gorilla, Macaca nemestrina, Pig, Rhesus monkey, Xenopus tropicalis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11007
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000050, mir-153

          Mature miRNA Sequence:

          UUGCAUAGUCACAAAAGUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Chicken gga-miR-153-3p MIMAT0001119
          Chrysemys picta cpi-miR-153-3p MIMAT0037788
          Dog cfa-miR-153 MIMAT0009840
          Gorilla gorilla ggo-miR-153 MIMAT0002274
          Macaca nemestrina mne-miR-153 MIMAT0002273
          Pig ssc-miR-153 MIMAT0002160
          View More Rows
          Rhesus monkey mml-miR-153-3p MIMAT0002271
          Xenopus tropicalis xtr-miR-153 MIMAT0003607

          Stem-loop Details



          Chicken

          Stem-loop ID gga-mir-153
          Stem-loop Accession # MI0001188
          Stem-loop Sequence
          AGCGGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001119
          miRBase ID: gga-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-153-1
          Stem-loop Accession # MI0029496
          Stem-loop Sequence
          CAGCUGCCAGUGCCAUUUUUGUGAUUUGCAGCUAGUAAUCUGGGUCCAGUUGCAUAGUCACAAAAGUGAGCAUUGGCAGCCGUGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037788
          miRBase ID: cpi-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Stem-loop ID cpi-mir-153-2
          Stem-loop Accession # MI0029497
          Stem-loop Sequence
          GCGGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037788
          miRBase ID: cpi-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-153
          Stem-loop Accession # MI0010335
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009840
          miRBase ID: cfa-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-153
          Stem-loop Accession # MI0002567
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002274
          miRBase ID: ggo-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-153-1
          Stem-loop Accession # MI0002565
          Stem-loop Sequence
          CUCACAGCUGCCAGUGUCAUUUUUGUGAUCUGCAGCUAGUAUUCUCACUCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGCAGGUGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002273
          miRBase ID: mne-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Stem-loop ID mne-mir-153-2
          Stem-loop Accession # MI0002568
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002273
          miRBase ID: mne-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-153
          Stem-loop Accession # MI0002454
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002160
          miRBase ID: ssc-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-153-1
          Stem-loop Accession # MI0002563
          Stem-loop Sequence
          CUCACAGCUGCCAGUGUCAUUUUUGUGAUCUGCAGCUAGUAUUCUCACUCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGCAGGUGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002271
          miRBase ID: mml-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Stem-loop ID mml-mir-153-2
          Stem-loop Accession # MI0002566
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002271
          miRBase ID: mml-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-153-1
          Stem-loop Accession # MI0004847
          Stem-loop Sequence
          CACGGCUGCCCUUGUCAUUUUUGUGAUUUGCAGCUUGUAAUUUUGGUCUCAGUUGCAUAGUCACAAAAGUGAUCAUGGGCAGGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003607
          miRBase ID: xtr-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Stem-loop ID xtr-mir-153-2
          Stem-loop Accession # MI0004940
          Stem-loop Sequence
          AGCAGUUGCCAGUGUCAUUUUUGUGACGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUUAUUGGAAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003607
          miRBase ID: xtr-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11007
          TaqMan™ MicroRNA Assay : 000476
          mirVana® miRNA inhibitor : MH11007
          mirVana® miRNA mimic : MC11007


          miRBase Alias:

          Human: hsa-miR-153 (v9.2)
          Rat: rno-miR-153 (v9.2)
          Gallus gallus: gga-miR-153 (v19)
          Macaca mulatta: mml-miR-153 (v19)
          Pongo pygmaeus: ppy-miR-153 (v14)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline