Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11007
          Assay Name: cfa-miR-153
          miRBase Accession Number: MI0010335
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Species: Chicken, Chrysemys picta, Dog, Gorilla gorilla, Macaca nemestrina, Pig, Rhesus monkey, Xenopus tropicalis
          Product Type: mirVana® miRNA mimic
          Assay ID MC11007

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000050, mir-153

          Mature miRNA Sequence:

          UUGCAUAGUCACAAAAGUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Chicken gga-miR-153-3p MIMAT0001119
          Chrysemys picta cpi-miR-153-3p MIMAT0037788
          Dog cfa-miR-153 MIMAT0009840
          Gorilla gorilla ggo-miR-153 MIMAT0002274
          Macaca nemestrina mne-miR-153 MIMAT0002273
          Pig ssc-miR-153 MIMAT0002160
          View More Rows
          Rhesus monkey mml-miR-153-3p MIMAT0002271
          Xenopus tropicalis xtr-miR-153 MIMAT0003607

          Stem-loop Details



          Chicken

          Stem-loop ID gga-mir-153
          Stem-loop Accession # MI0001188
          Stem-loop Sequence
          AGCGGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001119
          miRBase ID: gga-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-153-1
          Stem-loop Accession # MI0029496
          Stem-loop Sequence
          CAGCUGCCAGUGCCAUUUUUGUGAUUUGCAGCUAGUAAUCUGGGUCCAGUUGCAUAGUCACAAAAGUGAGCAUUGGCAGCCGUGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037788
          miRBase ID: cpi-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Stem-loop ID cpi-mir-153-2
          Stem-loop Accession # MI0029497
          Stem-loop Sequence
          GCGGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037788
          miRBase ID: cpi-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-153
          Stem-loop Accession # MI0010335
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009840
          miRBase ID: cfa-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-153
          Stem-loop Accession # MI0002567
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002274
          miRBase ID: ggo-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-153-1
          Stem-loop Accession # MI0002565
          Stem-loop Sequence
          CUCACAGCUGCCAGUGUCAUUUUUGUGAUCUGCAGCUAGUAUUCUCACUCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGCAGGUGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002273
          miRBase ID: mne-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Stem-loop ID mne-mir-153-2
          Stem-loop Accession # MI0002568
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002273
          miRBase ID: mne-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-153
          Stem-loop Accession # MI0002454
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002160
          miRBase ID: ssc-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-153-1
          Stem-loop Accession # MI0002563
          Stem-loop Sequence
          CUCACAGCUGCCAGUGUCAUUUUUGUGAUCUGCAGCUAGUAUUCUCACUCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGCAGGUGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002271
          miRBase ID: mml-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Stem-loop ID mml-mir-153-2
          Stem-loop Accession # MI0002566
          Stem-loop Sequence
          AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002271
          miRBase ID: mml-miR-153-3p
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-153-1
          Stem-loop Accession # MI0004847
          Stem-loop Sequence
          CACGGCUGCCCUUGUCAUUUUUGUGAUUUGCAGCUUGUAAUUUUGGUCUCAGUUGCAUAGUCACAAAAGUGAUCAUGGGCAGGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003607
          miRBase ID: xtr-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA

          Stem-loop ID xtr-mir-153-2
          Stem-loop Accession # MI0004940
          Stem-loop Sequence
          AGCAGUUGCCAGUGUCAUUUUUGUGACGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUUAUUGGAAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003607
          miRBase ID: xtr-miR-153
          Mature miRNA Sequence: UUGCAUAGUCACAAAAGUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11007
          Ambion® Pre-miR™ miRNA Precursor : PM11007
          TaqMan™ MicroRNA Assay : 000476
          mirVana® miRNA inhibitor : MH11007


          miRBase Alias:

          Gallus gallus: gga-miR-153 (v19)
          Macaca mulatta: mml-miR-153 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline