Search
Search

| Assay Name: | cfa-miR-153 |
|---|---|
| miRBase Accession Number: | MI0010335 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGA |
| Species: | Chicken, Chrysemys picta, Dog, Gorilla gorilla, Macaca nemestrina, Pig, Rhesus monkey, Xenopus tropicalis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-153-3p | MIMAT0001119 |
| Chrysemys picta | cpi-miR-153-3p | MIMAT0037788 |
| Dog | cfa-miR-153 | MIMAT0009840 |
| Gorilla gorilla | ggo-miR-153 | MIMAT0002274 |
| Macaca nemestrina | mne-miR-153 | MIMAT0002273 |
| Pig | ssc-miR-153 | MIMAT0002160 |
| View More Rows | ||
| Rhesus monkey | mml-miR-153-3p | MIMAT0002271 |
| Xenopus tropicalis | xtr-miR-153 | MIMAT0003607 |